Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478687_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-1285-3p
          Stem-loop Accession Number: MI0006346
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 92204015 - 92204098 [-] on Build GRCh38
          Mature miRNA Sequence: UCUGGGCAACAAAGUGAGACCU
          Species: Human, Pan troglodytes, Pongo pygmaeus
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478687_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000559, mir-1285

          Mature miRNA Sequence:

          UCUGGGCAACAAAGUGAGACCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-1285-3p MIMAT0005876
          Pan troglodytes ptr-miR-1285 MIMAT0008004
          Pongo pygmaeus ppy-miR-1285b MIMAT0016208

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-1285-1
          Stem-loop Accession # MI0006346
          Stem-loop Sequence
          UGUAGAGAUAGGAUCUCACUUUGUUGCCCAGGCUGGUCUCAAACUCCUGGUCUGGGCAACAAAGUGAGACCUUAUCUCUACAAG
          Chromosome Location Chr. 7 - 92204015 - 92204098 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005876
          miRBase ID: hsa-miR-1285-3p
          Mature miRNA Sequence: UCUGGGCAACAAAGUGAGACCU
          Chromosome Location: Chr. 7 - 92204015 - 92204098 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1285-2
          Stem-loop Accession # MI0006347
          Stem-loop Sequence
          UUUGGGAGGCCGAGGCUGGUGCAUCACUUGAGCCCAGCAAUUUGAGACCAAUCUGGGCAACAAAGUGAGACCUCCGUCUCUACAAAGA
          Chromosome Location Chr. 2 - 70252918 - 70253005 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005876
          miRBase ID: hsa-miR-1285-3p
          Mature miRNA Sequence: UCUGGGCAACAAAGUGAGACCU
          Chromosome Location: Chr. 7 - 92204015 - 92204098 [-] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-1285
          Stem-loop Accession # MI0008486
          Stem-loop Sequence
          UGUAGAGAUAGGAUCUCACUUUGUUGCCCAGGCUGGUCUCAAACUCCUGGUCUGGGCAACAAAGUGAGACCUUAUCUCUACAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008004
          miRBase ID: ptr-miR-1285
          Mature miRNA Sequence: UCUGGGCAACAAAGUGAGACCU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-1285b
          Stem-loop Accession # MI0015242
          Stem-loop Sequence
          UUUGGGAGGCCGAGGCUGGUGCAUCGCUUGAGCCUAGCAAUUUGAGACCAAUCUGGGCAACAAAGUGAGACCUCCAUCUCUACAAAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016208
          miRBase ID: ppy-miR-1285b
          Mature miRNA Sequence: UCUGGGCAACAAAGUGAGACCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305503_pri
          TaqMan™ Pri-miRNA Assay : Hs04226524_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13580
          Ambion® Pre-miR™ miRNA Precursor : PM13580
          TaqMan™ MicroRNA Assay : 002822
          mirVana® miRNA inhibitor : MH13580
          mirVana® miRNA mimic : MC13580
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 4
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human B Card
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 4

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline