Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13580
          Assay Name: hsa-miR-1285-3p
          miRBase Accession Number: MI0006346
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 92204015 - 92204098 [-] on Build GRCh38
          Mature miRNA Sequence: UCUGGGCAACAAAGUGAGACCU
          Species: Human, Pan troglodytes, Pongo pygmaeus
          Product Type: mirVana® miRNA mimic
          Assay ID MC13580

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000559, mir-1285

          Mature miRNA Sequence:

          UCUGGGCAACAAAGUGAGACCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-1285-3p MIMAT0005876
          Pan troglodytes ptr-miR-1285 MIMAT0008004
          Pongo pygmaeus ppy-miR-1285b MIMAT0016208

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-1285-1
          Stem-loop Accession # MI0006346
          Stem-loop Sequence
          UGUAGAGAUAGGAUCUCACUUUGUUGCCCAGGCUGGUCUCAAACUCCUGGUCUGGGCAACAAAGUGAGACCUUAUCUCUACAAG
          Chromosome Location Chr. 7 - 92204015 - 92204098 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005876
          miRBase ID: hsa-miR-1285-3p
          Mature miRNA Sequence: UCUGGGCAACAAAGUGAGACCU
          Chromosome Location: Chr. 7 - 92204015 - 92204098 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1285-2
          Stem-loop Accession # MI0006347
          Stem-loop Sequence
          UUUGGGAGGCCGAGGCUGGUGCAUCACUUGAGCCCAGCAAUUUGAGACCAAUCUGGGCAACAAAGUGAGACCUCCGUCUCUACAAAGA
          Chromosome Location Chr. 2 - 70252918 - 70253005 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005876
          miRBase ID: hsa-miR-1285-3p
          Mature miRNA Sequence: UCUGGGCAACAAAGUGAGACCU
          Chromosome Location: Chr. 7 - 92204015 - 92204098 [-] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-1285
          Stem-loop Accession # MI0008486
          Stem-loop Sequence
          UGUAGAGAUAGGAUCUCACUUUGUUGCCCAGGCUGGUCUCAAACUCCUGGUCUGGGCAACAAAGUGAGACCUUAUCUCUACAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008004
          miRBase ID: ptr-miR-1285
          Mature miRNA Sequence: UCUGGGCAACAAAGUGAGACCU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-1285b
          Stem-loop Accession # MI0015242
          Stem-loop Sequence
          UUUGGGAGGCCGAGGCUGGUGCAUCGCUUGAGCCUAGCAAUUUGAGACCAAUCUGGGCAACAAAGUGAGACCUCCAUCUCUACAAAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016208
          miRBase ID: ppy-miR-1285b
          Mature miRNA Sequence: UCUGGGCAACAAAGUGAGACCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305503_pri
          TaqMan™ Pri-miRNA Assay : Hs04226524_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13580
          Ambion® Pre-miR™ miRNA Precursor : PM13580
          TaqMan™ MicroRNA Assay : 002822
          mirVana® miRNA inhibitor : MH13580
          TaqMan™ Advanced miRNA Assay : 478687_mir


          miRBase Alias:

          Human: hsa-miR-1285 (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline