Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478971_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-512-3p
          Stem-loop Accession Number: MI0003140
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 53666679 - 53666762 [+] on Build GRCh38
          Mature miRNA Sequence: AAGUGCUGUCAUAGCUGAGGUC
          Species: Human, Pan troglodytes, Pongo pygmaeus
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478971_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000130, mir-506

          Mature miRNA Sequence:

          AAGUGCUGUCAUAGCUGAGGUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-512-3p MIMAT0002823
          Pan troglodytes ptr-miR-512 MIMAT0008180
          Pongo pygmaeus ppy-miR-512-3p MIMAT0015937

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-512-1
          Stem-loop Accession # MI0003140
          Stem-loop Sequence
          UCUCAGUCUGUGGCACUCAGCCUUGAGGGCACUUUCUGGUGCCAGAAUGAAAGUGCUGUCAUAGCUGAGGUCCAAUGACUGAGG
          Chromosome Location Chr. 19 - 53666679 - 53666762 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002823
          miRBase ID: hsa-miR-512-3p
          Mature miRNA Sequence: AAGUGCUGUCAUAGCUGAGGUC
          Chromosome Location: Chr. 19 - 53669157 - 53669254 [+] on Build GRCh38

          Stem-loop ID hsa-mir-512-2
          Stem-loop Accession # MI0003141
          Stem-loop Sequence
          GGUACUUCUCAGUCUGUGGCACUCAGCCUUGAGGGCACUUUCUGGUGCCAGAAUGAAAGUGCUGUCAUAGCUGAGGUCCAAUGACUGAGGCGAGCACC
          Chromosome Location Chr. 19 - 53669157 - 53669254 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002823
          miRBase ID: hsa-miR-512-3p
          Mature miRNA Sequence: AAGUGCUGUCAUAGCUGAGGUC
          Chromosome Location: Chr. 19 - 53669157 - 53669254 [+] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-512
          Stem-loop Accession # MI0008702
          Stem-loop Sequence
          CUCAGUCUGUGGCACUCAGCCUUGAGGGCACUUUCUGGUGUCAGAAUGAAAGUGCUGUCAUAGCUGAGGUCCAAUGACUGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008180
          miRBase ID: ptr-miR-512
          Mature miRNA Sequence: AAGUGCUGUCAUAGCUGAGGUC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-512-1
          Stem-loop Accession # MI0014988
          Stem-loop Sequence
          UCUCAGUCUGUGGCGCUCAGCCUUGGGGGCACUUUCUGGUGCCAGAAUGAAAGUGCUGUCAUAGCUGAGGUCCAAUGACUGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015937
          miRBase ID: ppy-miR-512-3p
          Mature miRNA Sequence: AAGUGCUGUCAUAGCUGAGGUC
          Chromosome Location: NA

          Stem-loop ID ppy-mir-512-2
          Stem-loop Accession # MI0014989
          Stem-loop Sequence
          GGUACUUCUCAGUCUGUGGCGCUCAGCCUUGGGGGCACUUUCUGGUGCCAGAAUGAAAGUGCUGUCAUAGCUGAGGUCCAAUGACUGAGGCGAGCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015937
          miRBase ID: ppy-miR-512-3p
          Mature miRNA Sequence: AAGUGCUGUCAUAGCUGAGGUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03295776_pri
          TaqMan™ Pri-miRNA Assay : Hs03295356_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10141
          Ambion® Pre-miR™ miRNA Precursor : PM10141
          TaqMan™ MicroRNA Assay : 001823
          mirVana® miRNA inhibitor : MH10141
          mirVana® miRNA mimic : MC10141
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 3
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 3
          MIRNA : TaqMan® Advanced miRNA Human A Card
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline