Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03295776_pri
          Assay Name:
          hsa-mir-512-2
          Stem-loop ID:
          hsa-mir-512-2
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.19: 53669157 - 53669254 [+] on Build GRCh38
          Stem-loop Location:
          Chr.19: 53669157 - 53669254 [+] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-512-5p hsa-miR-512-3p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs03295776_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          141

          Chromosome Location:

          Chr. 19 - 53669607 - 53669607 [-] on 53669607 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-512-5p MIMAT0002822
          Human hsa-miR-512-3p MIMAT0002823

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-512-2
          Stem-loop Accession # MI0003141
          Stem-loop Sequence
          GGUACUUCUCAGUCUGUGGCACUCAGCCUUGAGGGCACUUUCUGGUGCCAGAAUGAAAGUGCUGUCAUAGCUGAGGUCCAAUGACUGAGGCGAGCACC
          Chromosome Location Chr. 19 - 53669157 - 53669254 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002822
          miRBase ID: hsa-miR-512-5p
          Mature miRNA Sequence: CACUCAGCCUUGAGGGCACUUUC
          Chromosome Location: Chr. 19 - 53669157 - 53669254 [+] on Build GRCh38

          miRBase Accession #: MIMAT0002823
          miRBase ID: hsa-miR-512-3p
          Mature miRNA Sequence: AAGUGCUGUCAUAGCUGAGGUC
          Chromosome Location: Chr. 19 - 53669157 - 53669254 [+] on Build GRCh38

          Gene Family ID MIPF0000130, mir-506
          Distance to Amplicon 279 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10944
          Ambion® Pre-miR™ miRNA Precursor : PM10944
          TaqMan™ MicroRNA Assay : 001145
          mirVana® miRNA inhibitor : MH10944
          mirVana® miRNA mimic : MC10944
          TaqMan™ Advanced miRNA Assay : 478972_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10141
          Ambion® Pre-miR™ miRNA Precursor : PM10141
          TaqMan™ MicroRNA Assay : 001823
          mirVana® miRNA inhibitor : MH10141
          mirVana® miRNA mimic : MC10141
          TaqMan™ Advanced miRNA Assay : 478971_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline