Search
Search

| Assay Name: | hsa-miR-513b-5p |
|---|---|
| Stem-loop Accession Number: | MI0006648 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUCACAAGGAGGUGUCAUUUAU |
| Species: | Human, Pongo pygmaeus, Pygathrix bieti, Rhesus monkey, Symphalangus syndactylus |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | UUCACAAGGAGGUGUCAUUUAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-513b-5p | MIMAT0005788 |
| Pongo pygmaeus | ppy-miR-513b | MIMAT0015940 |
| Pygathrix bieti | pbi-miR-513b | MIMAT0005785 |
| Rhesus monkey | mml-miR-513b | MIMAT0005767 |
| Symphalangus syndactylus | ssy-miR-513b | MIMAT0005759 |