Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13754
          Assay Name: hsa-miR-513b-5p
          miRBase Accession Number: MI0006648
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 147199044 - 147199127 [-] on Build GRCh38
          Mature miRNA Sequence: UUCACAAGGAGGUGUCAUUUAU
          Species: Human, Pongo pygmaeus, Pygathrix bieti, Rhesus monkey, Symphalangus syndactylus
          Product Type: mirVana® miRNA mimic
          Assay ID MC13754

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000130, mir-506

          Mature miRNA Sequence:

          UUCACAAGGAGGUGUCAUUUAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-513b-5p MIMAT0005788
          Pongo pygmaeus ppy-miR-513b MIMAT0015940
          Pygathrix bieti pbi-miR-513b MIMAT0005785
          Rhesus monkey mml-miR-513b MIMAT0005767
          Symphalangus syndactylus ssy-miR-513b MIMAT0005759

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-513b
          Stem-loop Accession # MI0006648
          Stem-loop Sequence
          GUGUACAGUGCCUUUCACAAGGAGGUGUCAUUUAUGUGAACUAAAAUAUAAAUGUCACCUUUUUGAGAGGAGUAAUGUACAGCA
          Chromosome Location Chr. X - 147199044 - 147199127 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005788
          miRBase ID: hsa-miR-513b-5p
          Mature miRNA Sequence: UUCACAAGGAGGUGUCAUUUAU
          Chromosome Location: Chr. X - 147199044 - 147199127 [-] on Build GRCh38

          Pongo pygmaeus

          Stem-loop ID ppy-mir-513b
          Stem-loop Accession # MI0014992
          Stem-loop Sequence
          GUGUACAGUGCCUUUCACAAGGAGGUGUCAUUUAUGUGAACUAAAAUGUAAAUGUCACCUUUUUGAGAAGAGUAAUGUACAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015940
          miRBase ID: ppy-miR-513b
          Mature miRNA Sequence: UUCACAAGGAGGUGUCAUUUAU
          Chromosome Location: NA

          Pygathrix bieti

          Stem-loop ID pbi-mir-513b
          Stem-loop Accession # MI0006643
          Stem-loop Sequence
          UGUGCAGUGCCUUUCACAAGGAGGUGUCAUUUAUGUGAACUAAAGUAUAAAUGUCACCUUUUUGAGAAGAGUAAUGUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005785
          miRBase ID: pbi-miR-513b
          Mature miRNA Sequence: UUCACAAGGAGGUGUCAUUUAU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-513b-1
          Stem-loop Accession # MI0006616
          Stem-loop Sequence
          UUUACAGUGCCUUUCACAAGGAGGUGUCAUUUAUGUGAACUAAACUAUAAAUGUCACCUUUUUGGGAAGAGUAAUGUACAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005767
          miRBase ID: mml-miR-513b
          Mature miRNA Sequence: UUCACAAGGAGGUGUCAUUUAU
          Chromosome Location: NA

          Stem-loop ID mml-mir-513b-2
          Stem-loop Accession # MI0006617
          Stem-loop Sequence
          GUGUGCAGUGCCUUUCACAAGGAGGUGUCAUUUAUGUGAACUAAACUAUAAAUGUCACCUUUUUGGGAAGAGUAAUGUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005767
          miRBase ID: mml-miR-513b
          Mature miRNA Sequence: UUCACAAGGAGGUGUCAUUUAU
          Chromosome Location: NA

          Symphalangus syndactylus

          Stem-loop ID ssy-mir-513b-1
          Stem-loop Accession # MI0006606
          Stem-loop Sequence
          AGUGUACAGUGCCUUUCACAAGGAGGUGUCAUUUAUGUGAACUAAAAUAUAAAUGUGACCUUCUUGAGAAGAGUAAUGUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005759
          miRBase ID: ssy-miR-513b
          Mature miRNA Sequence: UUCACAAGGAGGUGUCAUUUAU
          Chromosome Location: NA

          Stem-loop ID ssy-mir-513b-2
          Stem-loop Accession # MI0006608
          Stem-loop Sequence
          UGUGCAGUGCCUUUCACAAGGAGGUGUCAUUUAUGUGAACUAAAAUAUAAAUUUCACCUUUUUGAGAAGGGUAAUGUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005759
          miRBase ID: ssy-miR-513b
          Mature miRNA Sequence: UUCACAAGGAGGUGUCAUUUAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03306086_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13754
          Ambion® Pre-miR™ miRNA Precursor : PM13754
          TaqMan™ MicroRNA Assay : 002757
          mirVana® miRNA inhibitor : MH13754
          TaqMan™ Advanced miRNA Assay : 479297_mir


          miRBase Alias:

          Human: hsa-miR-513b (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline