Search
Search

| Assay Name: | hsa-miR-3064-5p |
|---|---|
| Stem-loop Accession Number: | MI0017375 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCUGGCUGUUGUGGUGUGCAA |
| Species: | Human, Pteropus alecto |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0001238, mir-3064 |
Mature miRNA Sequence: | UCUGGCUGUUGUGGUGUGCAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3064-5p | MIMAT0019864 |
| Pteropus alecto | pal-miR-3064-5p | MIMAT0040404 |