Search
Search

Amplicon Length: | 91 |
Chromosome Location: | Chr. 17 - 64500511 - 64500511 [+] on 64500511 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3064-5p | MIMAT0019864 |
| Human | hsa-miR-3064-3p | MIMAT0019865 |
| Human | hsa-miR-5047 | MIMAT0020541 |