Search
Search

| Assay Name: | hsa-miR-133b |
|---|---|
| Stem-loop Accession Number: | MI0000822 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUGGUCCCCUUCAACCAGCUA |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Bovine, Columba livia, Dasypus novemcinctus, Dog, Guinea pig, Horse, Ornithorhynchus anatinus, Pan troglodytes, Pongo pygmaeus, Python bivittatus, Rabbit, Rhesus monkey, Zebrafish |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000029, mir-133 |
Mature miRNA Sequence: | UUUGGUCCCCUUCAACCAGCUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-133b | MIMAT0000770 |
| Mouse | mmu-miR-133b-3p | MIMAT0000769 |
| Rat | rno-miR-133b-3p | MIMAT0003126 |
| Alligator mississippiensis | ami-miR-133b-3p | MIMAT0038148 |
| Bovine | bta-miR-133b | MIMAT0009226 |
| Columba livia | cli-miR-133b-3p | MIMAT0038512 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-133b-3p | MIMAT0047636 |
| Dog | cfa-miR-133b | MIMAT0009835 |
| Guinea pig | cpo-miR-133b-3p | MIMAT0046989 |
| Horse | eca-miR-133b | MIMAT0013097 |
| Ornithorhynchus anatinus | oan-miR-133b-3p | MIMAT0006997 |
| Pan troglodytes | ptr-miR-133b | MIMAT0008030 |
| Pongo pygmaeus | ppy-miR-133b | MIMAT0015756 |
| Python bivittatus | pbv-miR-133b-3p | MIMAT0038939 |
| Rabbit | ocu-miR-133b-3p | MIMAT0048224 |
| Rhesus monkey | mml-miR-133b-3p | MIMAT0006189 |
| Zebrafish | dre-miR-133b-3p | MIMAT0001831 |