Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10029
          Assay Name: hsa-miR-133b
          miRBase Accession Number: MI0000822
          miRBase Version: v22.1
          Chromosome Location: Chr.6: 52148923 - 52149041 [+] on Build GRCh38
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Species: Human, Mouse, Rat, Alligator mississippiensis, Bovine, Columba livia, Dasypus novemcinctus, Dog, Guinea pig, Horse, Ornithorhynchus anatinus, Pan troglodytes, Pongo pygmaeus, Python bivittatus, Rabbit, Rhesus monkey, Zebrafish
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10029

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000029, mir-133

          Mature miRNA Sequence:

          UUUGGUCCCCUUCAACCAGCUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-133b MIMAT0000770
          Mouse mmu-miR-133b-3p MIMAT0000769
          Rat rno-miR-133b-3p MIMAT0003126
          Alligator mississippiensis ami-miR-133b-3p MIMAT0038148
          Bovine bta-miR-133b MIMAT0009226
          Columba livia cli-miR-133b-3p MIMAT0038512
          View More Rows
          Dasypus novemcinctus dno-miR-133b-3p MIMAT0047636
          Dog cfa-miR-133b MIMAT0009835
          Guinea pig cpo-miR-133b-3p MIMAT0046989
          Horse eca-miR-133b MIMAT0013097
          Ornithorhynchus anatinus oan-miR-133b-3p MIMAT0006997
          Pan troglodytes ptr-miR-133b MIMAT0008030
          Pongo pygmaeus ppy-miR-133b MIMAT0015756
          Python bivittatus pbv-miR-133b-3p MIMAT0038939
          Rabbit ocu-miR-133b-3p MIMAT0048224
          Rhesus monkey mml-miR-133b-3p MIMAT0006189
          Zebrafish dre-miR-133b-3p MIMAT0001831

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-133b
          Stem-loop Accession # MI0000822
          Stem-loop Sequence
          CCUCAGAAGAAAGAUGCCCCCUGCUCUGGCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUACAGCAGGGCUGGCAAUGCCCAGUCCUUGGAGA
          Chromosome Location Chr. 6 - 52148923 - 52149041 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000770
          miRBase ID: hsa-miR-133b
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: Chr. 6 - 52148923 - 52149041 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-133b
          Stem-loop Accession # MI0000821
          Stem-loop Sequence
          CCUCCAAAGGGAGUGGCCCCCUGCUCUGGCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUACAGCAGGGCUGGCAAAGCUCAAUAUUUGGAGA
          Chromosome Location Chr. 1 - 20682769 - 20682887 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000769
          miRBase ID: mmu-miR-133b-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: Chr. 1 - 20682769 - 20682887 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-133b
          Stem-loop Accession # MI0003490
          Stem-loop Sequence
          CCCUGCUCUGGCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUACAGCAGGGCUGGCAA
          Chromosome Location Chr. 9 - 26795649 - 26795732 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003126
          miRBase ID: rno-miR-133b-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: Chr. 9 - 26795649 - 26795732 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-133b
          Stem-loop Accession # MI0029736
          Stem-loop Sequence
          AGCUGCACUCUGCUGUGGCUGGUCAAACGGAACCAAGUCCGUCUUCCUUGGAGGUUUGGUCCCCUUCAACCAGCUAUAGCAGUGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038148
          miRBase ID: ami-miR-133b-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-133b
          Stem-loop Accession # MI0009734
          Stem-loop Sequence
          CCCUGCUCUGGCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUACAGCAGGGCUGGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009226
          miRBase ID: bta-miR-133b
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-133b
          Stem-loop Accession # MI0029966
          Stem-loop Sequence
          UAUGCUCUGCUCUGGCUGGUCAAACGGAACCAAGCCCAUCUUCCUCGGAGGUUUGGUCCCCUUCAACCAGCUAUAGCAGUGUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038512
          miRBase ID: cli-miR-133b-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-133b
          Stem-loop Accession # MI0038979
          Stem-loop Sequence
          GCUGGUCAAACGGAACCAAGUCCGUCUUCUUGAGAGGUUUGGUCCCCUUCAACCAGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047636
          miRBase ID: dno-miR-133b-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-133b
          Stem-loop Accession # MI0010327
          Stem-loop Sequence
          CCCUGCUCUGGCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUACAGCAGGGCUGGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009835
          miRBase ID: cfa-miR-133b
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-133b
          Stem-loop Accession # MI0038638
          Stem-loop Sequence
          GCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046989
          miRBase ID: cpo-miR-133b-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-133b
          Stem-loop Accession # MI0012844
          Stem-loop Sequence
          UGGCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013097
          miRBase ID: eca-miR-133b
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-133b
          Stem-loop Accession # MI0006780
          Stem-loop Sequence
          CGGCUGAACCCUGCUCUGGCUGGUCAAACGGAACCAAGUCCGUCUGCCUGAGAGGUUUGGUCCCCUUCAACCAGCUACAGCGGGGCCGACAUCGUCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006997
          miRBase ID: oan-miR-133b-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-133b
          Stem-loop Accession # MI0008534
          Stem-loop Sequence
          CUCAGAAGAAAGAUGCCCCCUGCUCUGGCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUACAGCAGGGCUGGCAAUGCCCAGUCCUUGGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008030
          miRBase ID: ptr-miR-133b
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-133b
          Stem-loop Accession # MI0014826
          Stem-loop Sequence
          CCUCAGAAGAAAGAUGCCCCCUGCUCUGGCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUACAGCAGGGCUGGCAAUGCCAAGUACUUGGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015756
          miRBase ID: ppy-miR-133b
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-133b
          Stem-loop Accession # MI0030230
          Stem-loop Sequence
          GAAGAAGAGCAUUGCUCUGGCUGGUCAAAAGGAACCAAGGCCGUCUUUCCUGGAGGUUUGGUCCCCUUCAACCAGCUAUAGCAGUGCUGACCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038939
          miRBase ID: pbv-miR-133b-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-133b
          Stem-loop Accession # MI0039288
          Stem-loop Sequence
          GCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048224
          miRBase ID: ocu-miR-133b-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-133b
          Stem-loop Accession # MI0007622
          Stem-loop Sequence
          CCUCAGAAGAAAGAUGCCCCCUGCUCUGGCUGGUCAAACGGAACCAAGUCCGUCUUCCUGAGAGGUUUGGUCCCCUUCAACCAGCUACAGCAGGGCUGGCAAUUCCCAGUCCUUGGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006189
          miRBase ID: mml-miR-133b-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-133b
          Stem-loop Accession # MI0001994
          Stem-loop Sequence
          CACGCCUUGCUGUGGCUGGUCAAAUGGAACCAAGUCAGGUGUUUCUGCGAGGUUUGGUCCCCUUCAACCAGCUACUGCGUCGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001831
          miRBase ID: dre-miR-133b-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303651_pri
          TaqMan™ Pri-miRNA Assay : Mm03307410_pri
          TaqMan™ Pri-miRNA Assay : Rn03465381_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10029
          Ambion® Pre-miR™ miRNA Precursor : PM10029
          TaqMan™ MicroRNA Assay : 002247
          mirVana® miRNA mimic : MC10029
          TaqMan™ Advanced miRNA Assay : 480871_mir


          miRBase Alias:

          Mouse: mmu-miR-133b (v17)
          Rat: rno-miR-133b (v18)
          Danio rerio: dre-miR-133b (v18)
          Macaca mulatta: mml-miR-133b (v19)
          Ochrobactrum anthropi: oan-miR-133b (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline