Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 483071_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-135a-2-3p
          Stem-loop Accession Number: MI0000453
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 97563812 - 97563911 [+] on Build GRCh38
          Mature miRNA Sequence: AUGUAGGGAUGGAAGCCAUGAA
          Species: Human, Alligator mississippiensis, Anolis carolinensis, Chicken, Columba livia, Monodelphis domestica, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 483071_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000028, mir-135

          Mature miRNA Sequence:

          AUGUAGGGAUGGAAGCCAUGAA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-135a-2-3p MIMAT0037309
          Alligator mississippiensis ami-miR-135-2-3p MIMAT0038151
          Anolis carolinensis aca-miR-135-2-3p MIMAT0021752
          Chicken gga-miR-135a-2-3p MIMAT0026489
          Columba livia cli-miR-135-2-3p MIMAT0038516
          Monodelphis domestica mdo-miR-135a-3p MIMAT0026666
          View More Rows
          Rhesus monkey mml-miR-135a-2-3p MIMAT0026567

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-135a-2
          Stem-loop Accession # MI0000453
          Stem-loop Sequence
          AGAUAAAUUCACUCUAGUGCUUUAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAAUACAUUGUGAAAAAUCA
          Chromosome Location Chr. 12 - 97563812 - 97563911 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0037309
          miRBase ID: hsa-miR-135a-2-3p
          Mature miRNA Sequence: AUGUAGGGAUGGAAGCCAUGAA
          Chromosome Location: Chr. 12 - 97563812 - 97563911 [+] on Build GRCh38

          Alligator mississippiensis

          Stem-loop ID ami-mir-135-2
          Stem-loop Accession # MI0029738
          Stem-loop Sequence
          UAAAUUCACUCUAGUGUUUUAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAAUACAUUGUGAAAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038151
          miRBase ID: ami-miR-135-2-3p
          Mature miRNA Sequence: AUGUAGGGAUGGAAGCCAUGAA
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-135-2
          Stem-loop Accession # MI0018742
          Stem-loop Sequence
          UAAAUUCACUCUAGUGUUUUAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAAUACAUUGUGAAAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021752
          miRBase ID: aca-miR-135-2-3p
          Mature miRNA Sequence: AUGUAGGGAUGGAAGCCAUGAA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-135a-2
          Stem-loop Accession # MI0001169
          Stem-loop Sequence
          AGAUAAAUUCACUCUAGUGUUUUAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAAUACAUUGUGAAAAAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026489
          miRBase ID: gga-miR-135a-2-3p
          Mature miRNA Sequence: AUGUAGGGAUGGAAGCCAUGAA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-135-2
          Stem-loop Accession # MI0029969
          Stem-loop Sequence
          CUAGUGUUUUAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAAUACAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038516
          miRBase ID: cli-miR-135-2-3p
          Mature miRNA Sequence: AUGUAGGGAUGGAAGCCAUGAA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-135a
          Stem-loop Accession # MI0005297
          Stem-loop Sequence
          AGAUAAAUUCACUCUAGUGUCUUAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAAUACAUUGUGAAAUGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026666
          miRBase ID: mdo-miR-135a-3p
          Mature miRNA Sequence: AUGUAGGGAUGGAAGCCAUGAA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-135a-2
          Stem-loop Accession # MI0002540
          Stem-loop Sequence
          AGAUAAAUUCACUCUAGUGCUUUAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAAUACAUUGUGAAAAAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026567
          miRBase ID: mml-miR-135a-2-3p
          Mature miRNA Sequence: AUGUAGGGAUGGAAGCCAUGAA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303133_pri
          TaqMan™ MicroRNA Assay : 477768_mat
          mirVana® miRNA inhibitor : MH23887
          mirVana® miRNA mimic : MC23887

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline