Search
Search

| Assay Name: | hsa-miR-23b-3p |
|---|---|
| Stem-loop Accession Number: | MI0000439 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUCACAUUGCCAGGGAUUACCAC |
| Species: | Human, Alligator mississippiensis, Bovine, Dasypus novemcinctus, Guinea pig, Ophiophagus hannah, Oryzias latipes, Pan paniscus, Pan troglodytes, Petromyzon marinus, Pongo pygmaeus, Python bivittatus, Rabbit, Zebrafish |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000027, mir-23 |
Mature miRNA Sequence: | AUCACAUUGCCAGGGAUUACCAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-23b-3p | MIMAT0000418 |
| Alligator mississippiensis | ami-miR-23b-3p | MIMAT0038060 |
| Bovine | bta-miR-23b-3p | MIMAT0003852 |
| Dasypus novemcinctus | dno-miR-23b-3p | MIMAT0047531 |
| Guinea pig | cpo-miR-23b-3p | MIMAT0046879 |
| Ophiophagus hannah | oha-miR-23b-3p | MIMAT0036857 |
| View More Rows | ||
| Oryzias latipes | ola-miR-23b | MIMAT0022588 |
| Pan paniscus | ppa-miR-23b | MIMAT0002212 |
| Pan troglodytes | ptr-miR-23b | MIMAT0002210 |
| Petromyzon marinus | pma-miR-23b | MIMAT0019404 |
| Pongo pygmaeus | ppy-miR-23b | MIMAT0002211 |
| Python bivittatus | pbv-miR-23b-3p | MIMAT0038855 |
| Rabbit | ocu-miR-23b-3p | MIMAT0048129 |
| Zebrafish | dre-miR-23b-3p | MIMAT0048661 |