Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10499
          Assay Name: hsa-miR-23b-3p
          miRBase Accession Number: MI0000439
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 95085208 - 95085304 [+] on Build GRCh38
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Species: Human, Alligator mississippiensis, Bovine, Dasypus novemcinctus, Guinea pig, Ophiophagus hannah, Oryzias latipes, Pan paniscus, Pan troglodytes, Petromyzon marinus, Pongo pygmaeus, Python bivittatus, Rabbit, Zebrafish
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10499

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000027, mir-23

          Mature miRNA Sequence:

          AUCACAUUGCCAGGGAUUACCAC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-23b-3p MIMAT0000418
          Alligator mississippiensis ami-miR-23b-3p MIMAT0038060
          Bovine bta-miR-23b-3p MIMAT0003852
          Dasypus novemcinctus dno-miR-23b-3p MIMAT0047531
          Guinea pig cpo-miR-23b-3p MIMAT0046879
          Ophiophagus hannah oha-miR-23b-3p MIMAT0036857
          View More Rows
          Oryzias latipes ola-miR-23b MIMAT0022588
          Pan paniscus ppa-miR-23b MIMAT0002212
          Pan troglodytes ptr-miR-23b MIMAT0002210
          Petromyzon marinus pma-miR-23b MIMAT0019404
          Pongo pygmaeus ppy-miR-23b MIMAT0002211
          Python bivittatus pbv-miR-23b-3p MIMAT0038855
          Rabbit ocu-miR-23b-3p MIMAT0048129
          Zebrafish dre-miR-23b-3p MIMAT0048661

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-23b
          Stem-loop Accession # MI0000439
          Stem-loop Sequence
          CUCAGGUGCUCUGGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACCACGACCUUGGC
          Chromosome Location Chr. 9 - 95085208 - 95085304 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000418
          miRBase ID: hsa-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: Chr. 9 - 95085208 - 95085304 [+] on Build GRCh38

          Alligator mississippiensis

          Stem-loop ID ami-mir-23b
          Stem-loop Accession # MI0029679
          Stem-loop Sequence
          CCCAGUGUUGUGGCUGUUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAAAUUAAAAUCACAUUGCCAGGGAUUACCACAGAGCCAUGACCAUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038060
          miRBase ID: ami-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-23b
          Stem-loop Accession # MI0005066
          Stem-loop Sequence
          GGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003852
          miRBase ID: bta-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-23b
          Stem-loop Accession # MI0038920
          Stem-loop Sequence
          GGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047531
          miRBase ID: dno-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-23b
          Stem-loop Accession # MI0038576
          Stem-loop Sequence
          GGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046879
          miRBase ID: cpo-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-23b
          Stem-loop Accession # MI0031431
          Stem-loop Sequence
          UUUUGGUUGUUUGGGUUCCUGGCAUGCUGAUUUGUGAUUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACAUAACCGUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036857
          miRBase ID: oha-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA

          Oryzias latipes

          Stem-loop ID ola-mir-23b
          Stem-loop Accession # MI0019468
          Stem-loop Sequence
          UUUCAUGUCUGGGCUGUGAGGGUUCCUGGCGUGCUGAUUUGUGACUUAUGAUAAAAUCACAUUGCCAGGGAUUACCACACAGCCUUGACCCGGACGGCUGCUAUGGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022588
          miRBase ID: ola-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-23b
          Stem-loop Accession # MI0002501
          Stem-loop Sequence
          CUCAGGUGCUCUGGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACCACGACCUUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002212
          miRBase ID: ppa-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-23b
          Stem-loop Accession # MI0002499
          Stem-loop Sequence
          CUCAGGUGCUCUGGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACCACGACCUUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002210
          miRBase ID: ptr-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA

          Petromyzon marinus

          Stem-loop ID pma-mir-23b
          Stem-loop Accession # MI0017027
          Stem-loop Sequence
          CGAGGGCCGGAGGGUUCCUGGCAAGGUGAUUUGCGACUGCGUUUCUAAGAAAAAUCACAUUGCCAGGGAUUACCACUGGCCCUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019404
          miRBase ID: pma-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-23b
          Stem-loop Accession # MI0002500
          Stem-loop Sequence
          CUCAGGUGCUCUGGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACCACGACCUUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002211
          miRBase ID: ppy-miR-23b
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-23b
          Stem-loop Accession # MI0030171
          Stem-loop Sequence
          CUGUUUUGGUUGUUUGGGUUCCUGGCAUGCUGAUUUGUGAUUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACAUAACCAUGACAUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038855
          miRBase ID: pbv-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-23b
          Stem-loop Accession # MI0039235
          Stem-loop Sequence
          GGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048129
          miRBase ID: ocu-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-23b-2
          Stem-loop Accession # MI0039517
          Stem-loop Sequence
          CUGCUGGCUGUGUGGGUUCCUGGCGUGCUGAUUUGUGACUUAAGAUAAAAAUCACAUUGCCAGGGAUUACCACACAACCGCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048661
          miRBase ID: dre-miR-23b-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUACCAC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303058_pri
          TaqMan™ MicroRNA Assay : 245306_mat
          mirVana® miRNA mimic : MC10499
          TaqMan™ Advanced miRNA Assay : 483150_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline