Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10490
          Assay Name: hsa-miR-497-5p
          miRBase Accession Number: MI0003138
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 7017911 - 7018022 [-] on Build GRCh38
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Species: Human, Callithrix jacchus, Dog, Horse, Monodelphis domestica, Nomascus leucogenys, Otolemur garnettii, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Sarcophilus harrisii
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10490
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000231, mir-497

          Mature miRNA Sequence:

          CAGCAGCACACUGUGGUUUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-497-5p MIMAT0002820
          Callithrix jacchus cja-miR-497 MIMAT0039538
          Dog cfa-miR-497 MIMAT0006691
          Horse eca-miR-497 MIMAT0013043
          Monodelphis domestica mdo-miR-497-5p MIMAT0012762
          Nomascus leucogenys nle-miR-497 MIMAT0049381
          View More Rows
          Otolemur garnettii oga-miR-497 MIMAT0049634
          Pig ssc-miR-497 MIMAT0013926
          Pongo pygmaeus ppy-miR-497 MIMAT0015918
          Pteropus alecto pal-miR-497-5p MIMAT0040077
          Rhesus monkey mml-miR-497-5p MIMAT0006359
          Sarcophilus harrisii sha-miR-497 MIMAT0022821

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-497
          Stem-loop Accession # MI0003138
          Stem-loop Sequence
          CCACCCCGGUCCUGCUCCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGGGGAGGCACCGCCGAGG
          Chromosome Location Chr. 17 - 7017911 - 7018022 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002820
          miRBase ID: hsa-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: Chr. 17 - 7017911 - 7018022 [-] on Build GRCh38

          Callithrix jacchus

          Stem-loop ID cja-mir-497
          Stem-loop Accession # MI0032068
          Stem-loop Sequence
          CUCAACCCACCCGGUCCUGCUCCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGGGGAGGCACCGCCGAGGUUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039538
          miRBase ID: cja-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-497
          Stem-loop Accession # MI0008104
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGUACUGUGGCCACGUCCAGACCACACUGUGGUGUUAGGGUGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006691
          miRBase ID: cfa-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-497
          Stem-loop Accession # MI0012787
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013043
          miRBase ID: eca-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-497
          Stem-loop Accession # MI0012519
          Stem-loop Sequence
          CCUGCCUCCGCCCCAGCAGCACACUGUGGUUUGUUCAGUGUUAUGGCCAAGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012762
          miRBase ID: mdo-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-497
          Stem-loop Accession # MI0040119
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049381
          miRBase ID: nle-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-497
          Stem-loop Accession # MI0040417
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049634
          miRBase ID: oga-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-497
          Stem-loop Accession # MI0013139
          Stem-loop Sequence
          CCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013926
          miRBase ID: ssc-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-497
          Stem-loop Accession # MI0014970
          Stem-loop Sequence
          CCACCCCGGUCCUGCUCCCGCCCCAGCAGCACACUGUGGUUUGUGCGGCAUUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGGGGAGGCACCGCCGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015918
          miRBase ID: ppy-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-497
          Stem-loop Accession # MI0032482
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040077
          miRBase ID: pal-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-497
          Stem-loop Accession # MI0007771
          Stem-loop Sequence
          CCACCCCGGUCCUGCUCCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGGGGAGGCACCGCCGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006359
          miRBase ID: mml-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Sarcophilus harrisii

          Stem-loop ID sha-mir-497
          Stem-loop Accession # MI0019649
          Stem-loop Sequence
          CUACUCCCAAAUCCCAUCCUGCCUCCGCCCCAGCAGCACACUGUGGUUUGUUCAGUGUUGUGGCCAAGUCCAAACCACACUGUGGUGUUAGAGUGAGGGUAAGGAAGGCAUCACUGCCACCACCACAGCUUGGACCUGGAGCAGGACACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022821
          miRBase ID: sha-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303903_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10490
          TaqMan™ MicroRNA Assay : 001043
          mirVana® miRNA inhibitor : MH10490
          mirVana® miRNA mimic : MC10490
          TaqMan™ Advanced miRNA Assay : 478138_mir


          miRBase Alias:

          Human: hsa-miR-497 (v17)
          Macaca mulatta: mml-miR-497 (v19)
          Monodelphis domestica: mdo-miR-497 (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-6qggw:80/100.66.76.149:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline