Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10490
          Assay Name: hsa-miR-497-5p
          miRBase Accession Number: MI0003138
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 7017911 - 7018022 [-] on Build GRCh38
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Species: Human, Callithrix jacchus, Dog, Horse, Monodelphis domestica, Nomascus leucogenys, Otolemur garnettii, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Sarcophilus harrisii
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10490

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000231, mir-497

          Mature miRNA Sequence:

          CAGCAGCACACUGUGGUUUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-497-5p MIMAT0002820
          Callithrix jacchus cja-miR-497 MIMAT0039538
          Dog cfa-miR-497 MIMAT0006691
          Horse eca-miR-497 MIMAT0013043
          Monodelphis domestica mdo-miR-497-5p MIMAT0012762
          Nomascus leucogenys nle-miR-497 MIMAT0049381
          View More Rows
          Otolemur garnettii oga-miR-497 MIMAT0049634
          Pig ssc-miR-497 MIMAT0013926
          Pongo pygmaeus ppy-miR-497 MIMAT0015918
          Pteropus alecto pal-miR-497-5p MIMAT0040077
          Rhesus monkey mml-miR-497-5p MIMAT0006359
          Sarcophilus harrisii sha-miR-497 MIMAT0022821

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-497
          Stem-loop Accession # MI0003138
          Stem-loop Sequence
          CCACCCCGGUCCUGCUCCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGGGGAGGCACCGCCGAGG
          Chromosome Location Chr. 17 - 7017911 - 7018022 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002820
          miRBase ID: hsa-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: Chr. 17 - 7017911 - 7018022 [-] on Build GRCh38

          Callithrix jacchus

          Stem-loop ID cja-mir-497
          Stem-loop Accession # MI0032068
          Stem-loop Sequence
          CUCAACCCACCCGGUCCUGCUCCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGGGGAGGCACCGCCGAGGUUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039538
          miRBase ID: cja-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-497
          Stem-loop Accession # MI0008104
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGUACUGUGGCCACGUCCAGACCACACUGUGGUGUUAGGGUGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006691
          miRBase ID: cfa-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-497
          Stem-loop Accession # MI0012787
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013043
          miRBase ID: eca-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-497
          Stem-loop Accession # MI0012519
          Stem-loop Sequence
          CCUGCCUCCGCCCCAGCAGCACACUGUGGUUUGUUCAGUGUUAUGGCCAAGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012762
          miRBase ID: mdo-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-497
          Stem-loop Accession # MI0040119
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049381
          miRBase ID: nle-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-497
          Stem-loop Accession # MI0040417
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049634
          miRBase ID: oga-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-497
          Stem-loop Accession # MI0013139
          Stem-loop Sequence
          CCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013926
          miRBase ID: ssc-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-497
          Stem-loop Accession # MI0014970
          Stem-loop Sequence
          CCACCCCGGUCCUGCUCCCGCCCCAGCAGCACACUGUGGUUUGUGCGGCAUUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGGGGAGGCACCGCCGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015918
          miRBase ID: ppy-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-497
          Stem-loop Accession # MI0032482
          Stem-loop Sequence
          CAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040077
          miRBase ID: pal-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-497
          Stem-loop Accession # MI0007771
          Stem-loop Sequence
          CCACCCCGGUCCUGCUCCCGCCCCAGCAGCACACUGUGGUUUGUACGGCACUGUGGCCACGUCCAAACCACACUGUGGUGUUAGAGCGAGGGUGGGGGAGGCACCGCCGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006359
          miRBase ID: mml-miR-497-5p
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA

          Sarcophilus harrisii

          Stem-loop ID sha-mir-497
          Stem-loop Accession # MI0019649
          Stem-loop Sequence
          CUACUCCCAAAUCCCAUCCUGCCUCCGCCCCAGCAGCACACUGUGGUUUGUUCAGUGUUGUGGCCAAGUCCAAACCACACUGUGGUGUUAGAGUGAGGGUAAGGAAGGCAUCACUGCCACCACCACAGCUUGGACCUGGAGCAGGACACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022821
          miRBase ID: sha-miR-497
          Mature miRNA Sequence: CAGCAGCACACUGUGGUUUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303903_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10490
          Ambion® Pre-miR™ miRNA Precursor : PM10490
          TaqMan™ MicroRNA Assay : 001043
          mirVana® miRNA mimic : MC10490
          TaqMan™ Advanced miRNA Assay : 478138_mir


          miRBase Alias:

          Human: hsa-miR-497 (v17)
          Macaca mulatta: mml-miR-497 (v19)
          Monodelphis domestica: mdo-miR-497 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-v4tl5:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline