Search
Search

| Assay Name: | mmu-miR-465a-3p |
|---|---|
| miRBase Accession Number: | MI0002400 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAUCAGGGCCUUUCUAAGUAGA |
| Species: | Mouse |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000384, mir-465 |
Mature miRNA Sequence: | GAUCAGGGCCUUUCUAAGUAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-465a-3p | MIMAT0004217 |
| Mouse | mmu-miR-465b-3p | MIMAT0004872 |
| Mouse | mmu-miR-465c-3p | MIMAT0004874 |