Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03307546_pri
          Assay Name:
          mmu-mir-465a
          Stem-loop ID:
          mmu-mir-465a mmu-mir-465b-2
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.X: 66839052 - 66839125 [-] on Build GRCm38
          Stem-loop Location:
          Chr.X: 66839052 - 66839125 [-] on Build GRCm38 Chr.X: 66835764 - 66835842 [-] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-465a-3p mmu-miR-465a-5p mmu-miR-465b-3p mmu-miR-465b-5p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03307546_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          93

          Chromosome Location:

          Chr. X - 66838850 - 66838850 [+] on 66838850 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-465a-3p MIMAT0004217
          Mouse mmu-miR-465a-5p MIMAT0002106
          Mouse mmu-miR-465b-3p MIMAT0004872
          Mouse mmu-miR-465b-5p MIMAT0004871

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-465a
          Stem-loop Accession # MI0002400
          Stem-loop Sequence
          GCCCUAUUUAGAAUGGCACUGAUGUGAUAAAAUAAAAAAUUGAUCAGGGCCUUUCUAAGUAGAGUAAGGCUUAC
          Chromosome Location Chr. X - 66839052 - 66839125 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004217
          miRBase ID: mmu-miR-465a-3p
          Mature miRNA Sequence: GAUCAGGGCCUUUCUAAGUAGA
          Chromosome Location: Chr. X - 66839052 - 66839125 [-] on Build GRCm38

          miRBase Accession #: MIMAT0002106
          miRBase ID: mmu-miR-465a-5p
          Mature miRNA Sequence: UAUUUAGAAUGGCACUGAUGUGA
          Chromosome Location: Chr. X - 66839052 - 66839125 [-] on Build GRCm38

          Gene Family ID MIPF0000384, mir-465
          Distance to Amplicon 160 bases
          Stem-loop ID mmu-mir-465b-2
          Stem-loop Accession # MI0005499
          Stem-loop Sequence
          UGCAAUGCCCUAUUUAGAAUGGUGCUGAUCUGAUGAAAUAAAAAAUUGAUCAGGGCCUUUCUAAGUAGAGUAAGGCUUA
          Chromosome Location Chr. X - 66835764 - 66835842 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004872
          miRBase ID: mmu-miR-465b-3p
          Mature miRNA Sequence: GAUCAGGGCCUUUCUAAGUAGA
          Chromosome Location: Chr. X - 66835764 - 66835842 [-] on Build GRCm38

          miRBase Accession #: MIMAT0004871
          miRBase ID: mmu-miR-465b-5p
          Mature miRNA Sequence: UAUUUAGAAUGGUGCUGAUCUG
          Chromosome Location: Chr. X - 66835764 - 66835842 [-] on Build GRCm38

          Gene Family ID MIPF0000384, mir-465
          Distance to Amplicon 2958 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM12104
          Ambion® Pre-miR™ miRNA Precursor : PM12104
          TaqMan™ MicroRNA Assay : 002040
          mirVana® miRNA inhibitor : MH12104
          mirVana® miRNA mimic : MC12104
          TaqMan™ Advanced miRNA Assay : mmu482768_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM13014
          Ambion® Pre-miR™ miRNA Precursor : PM13014
          TaqMan™ MicroRNA Assay : 002485
          mirVana® miRNA inhibitor : MH13014
          mirVana® miRNA mimic : MC13014
          Ambion® Anti-miR™ miRNA Inhibitor : AM10752
          Ambion® Pre-miR™ miRNA Precursor : PM10752
          TaqMan™ MicroRNA Assay : 001082
          mirVana® miRNA inhibitor : MH10752
          mirVana® miRNA mimic : MC10752

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline