Search
Search

Amplicon Length: | 90 |
Chromosome Location: | Chr. X - 66822377 - 66822377 [-] on 66822377 Build GRCm38 |
Species: | Mouse |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-465c-5p | MIMAT0004873 |
| Mouse | mmu-miR-465c-3p | MIMAT0004874 |
| Mouse | mmu-miR-465d-5p | MIMAT0029880 |
| Mouse | mmu-miR-465d-3p | MIMAT0029881 |