Search
Search

| Assay Name: | hsa-miR-152-3p |
|---|---|
| miRBase Accession Number: | MI0000462 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCAGUGCAUGACAGAACUUGG |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Callithrix jacchus, Chrysemys picta, Cricetulus griseus, Dog, Gorilla gorilla, Guinea pig, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Sheep, Tupaia chinensis |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000056, mir-148 |
Mature miRNA Sequence: | UCAGUGCAUGACAGAACUUGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-152-3p | MIMAT0000438 |
| Mouse | mmu-miR-152-3p | MIMAT0000162 |
| Rat | rno-miR-152-3p | MIMAT0000854 |
| Alligator mississippiensis | ami-miR-152-3p | MIMAT0038183 |
| Callithrix jacchus | cja-miR-152 | MIMAT0039463 |
| Chrysemys picta | cpi-miR-152-3p | MIMAT0037786 |
| View More Rows | ||
| Cricetulus griseus | cgr-miR-152-3p | MIMAT0023785 |
| Dog | cfa-miR-152 | MIMAT0006738 |
| Gorilla gorilla | ggo-miR-152 | MIMAT0024099 |
| Guinea pig | cpo-miR-152-3p | MIMAT0047033 |
| Pan troglodytes | ptr-miR-152 | MIMAT0008046 |
| Pig | ssc-miR-152 | MIMAT0013887 |
| Pongo pygmaeus | ppy-miR-152 | MIMAT0024356 |
| Pteropus alecto | pal-miR-152-3p | MIMAT0040107 |
| Rabbit | ocu-miR-152-3p | MIMAT0048268 |
| Rhesus monkey | mml-miR-152-3p | MIMAT0006214 |
| Sheep | oar-miR-152 | MIMAT0030035 |
| Tupaia chinensis | tch-miR-152 | MIMAT0036545 |