Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481445_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-152-3p
          Stem-loop Accession Number: MI0000462
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 48037161 - 48037247 [-] on Build GRCh38
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Callithrix jacchus, Chrysemys picta, Cricetulus griseus, Dog, Gorilla gorilla, Guinea pig, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Sheep, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481445_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000056, mir-148

          Mature miRNA Sequence:

          UCAGUGCAUGACAGAACUUGG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-152-3p MIMAT0000438
          Mouse mmu-miR-152-3p MIMAT0000162
          Rat rno-miR-152-3p MIMAT0000854
          Alligator mississippiensis ami-miR-152-3p MIMAT0038183
          Callithrix jacchus cja-miR-152 MIMAT0039463
          Chrysemys picta cpi-miR-152-3p MIMAT0037786
          View More Rows
          Cricetulus griseus cgr-miR-152-3p MIMAT0023785
          Dog cfa-miR-152 MIMAT0006738
          Gorilla gorilla ggo-miR-152 MIMAT0024099
          Guinea pig cpo-miR-152-3p MIMAT0047033
          Pan troglodytes ptr-miR-152 MIMAT0008046
          Pig ssc-miR-152 MIMAT0013887
          Pongo pygmaeus ppy-miR-152 MIMAT0024356
          Pteropus alecto pal-miR-152-3p MIMAT0040107
          Rabbit ocu-miR-152-3p MIMAT0048268
          Rhesus monkey mml-miR-152-3p MIMAT0006214
          Sheep oar-miR-152 MIMAT0030035
          Tupaia chinensis tch-miR-152 MIMAT0036545

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-152
          Stem-loop Accession # MI0000462
          Stem-loop Sequence
          UGUCCCCCCCGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGAAGGACC
          Chromosome Location Chr. 17 - 48037161 - 48037247 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000438
          miRBase ID: hsa-miR-152-3p
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: Chr. 17 - 48037161 - 48037247 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-152
          Stem-loop Accession # MI0000174
          Stem-loop Sequence
          CCGGGCCUAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGG
          Chromosome Location Chr. 11 - 96850393 - 96850465 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000162
          miRBase ID: mmu-miR-152-3p
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: Chr. 11 - 96850393 - 96850465 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-152
          Stem-loop Accession # MI0000921
          Stem-loop Sequence
          UGUUCCCCGGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGUAGGAC
          Chromosome Location Chr. 10 - 84719319 - 84719403 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000854
          miRBase ID: rno-miR-152-3p
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: Chr. 10 - 84719319 - 84719403 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-152
          Stem-loop Accession # MI0029757
          Stem-loop Sequence
          GUCCUCUCAGCUCAGGUUCUGUGGUACACUUGGACUCGGACUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGUUUGGAUGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038183
          miRBase ID: ami-miR-152-3p
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-152
          Stem-loop Accession # MI0031987
          Stem-loop Sequence
          CCCGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGUAGUCAGUGCAUGACAGAACUUGGGCCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039463
          miRBase ID: cja-miR-152
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-152
          Stem-loop Accession # MI0029495
          Stem-loop Sequence
          GUCCUCUCGGCCCAGGUUCUGUGGUACACUUGGACUUGGACUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGUUUGGAUGGACCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037786
          miRBase ID: cpi-miR-152-3p
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-152
          Stem-loop Accession # MI0020413
          Stem-loop Sequence
          CCCGGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023785
          miRBase ID: cgr-miR-152-3p
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-152
          Stem-loop Accession # MI0008162
          Stem-loop Sequence
          AGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006738
          miRBase ID: cfa-miR-152
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-152
          Stem-loop Accession # MI0020646
          Stem-loop Sequence
          GAGGAGGCUCGGCCCGCUGUCCCCCCCGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGAAGGACCUUCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024099
          miRBase ID: ggo-miR-152
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-152
          Stem-loop Accession # MI0038661
          Stem-loop Sequence
          AGGUUCUGUGAUACACUCAGACUCGGACUCUGGAGCAGUCAGUGCAUGACAGAACUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047033
          miRBase ID: cpo-miR-152-3p
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-152
          Stem-loop Accession # MI0008552
          Stem-loop Sequence
          GUCCCCCCCGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGAAGGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008046
          miRBase ID: ptr-miR-152
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-152
          Stem-loop Accession # MI0013104
          Stem-loop Sequence
          CGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGACGGACCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013887
          miRBase ID: ssc-miR-152
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-152
          Stem-loop Accession # MI0020904
          Stem-loop Sequence
          GGAGGCUCGGCCCGCUGUCCCCCCCGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGAAGGACCUUCUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024356
          miRBase ID: ppy-miR-152
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-152
          Stem-loop Accession # MI0032497
          Stem-loop Sequence
          AGGUUCUGUGAUACACUCCGACUCGAGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040107
          miRBase ID: pal-miR-152-3p
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-152
          Stem-loop Accession # MI0039311
          Stem-loop Sequence
          AGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048268
          miRBase ID: ocu-miR-152-3p
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-152
          Stem-loop Accession # MI0007643
          Stem-loop Sequence
          UGUCCCCCCCGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGAAGGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006214
          miRBase ID: mml-miR-152-3p
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-152
          Stem-loop Accession # MI0025256
          Stem-loop Sequence
          GGCCCAGUUUCUGUGAUACUCUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGACGGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030035
          miRBase ID: oar-miR-152
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-152
          Stem-loop Accession # MI0031205
          Stem-loop Sequence
          AGGUUCUGUGAUACACUCAGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036545
          miRBase ID: tch-miR-152
          Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303177_pri
          TaqMan™ Pri-miRNA Assay : Rn03465082_pri
          TaqMan™ Pri-miRNA Assay : Mm03306379_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12269
          Ambion® Pre-miR™ miRNA Precursor : PM12269
          TaqMan™ MicroRNA Assay : 000475
          mirVana® miRNA inhibitor : MH12269
          mirVana® miRNA mimic : MC12269
          TaqMan™ Advanced miRNA Assay : 477921_mir
          TaqMan™ Advanced miRNA Assay : mmu482952_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline