Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm03306379_pri
Assay Name:
mmu-mir-152
Stem-loop ID:
mmu-mir-152
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.11: 96850393 - 96850465 [+] on Build GRCm38
Stem-loop Location:
Chr.11: 96850393 - 96850465 [+] on Build GRCm38
Mature miRNA ID:
mmu-miR-152-3p
mmu-miR-152-5p
Species:
Mouse
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Mm03306379_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
56
Chromosome Location:
Chr. 11 - 96850194 - 96850194 [-] on 96850194 Build GRCm38
Species:
Mouse
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Mouse
mmu-miR-152-3p
MIMAT0000162
Mouse
mmu-miR-152-5p
MIMAT0016991
Stem-loop Details
Mouse
Stem-loop ID
mmu-mir-152
Stem-loop Accession #
MI0000174
Stem-loop Sequence
CCGGGCCUAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGG
Chromosome Location
Chr. 11 - 96850393 - 96850465 [+] on Build GRCm38
Mature MicroRNA
miRBase Accession #: MIMAT0000162
miRBase ID: mmu-miR-152-3p
Mature miRNA Sequence: UCAGUGCAUGACAGAACUUGG
Chromosome Location: Chr. 11 - 96850393 - 96850465 [+] on Build GRCm38
miRBase Accession #: MIMAT0016991
miRBase ID: mmu-miR-152-5p
Mature miRNA Sequence: UAGGUUCUGUGAUACACUCCGACU
Chromosome Location: Chr. 11 - 96850393 - 96850465 [+] on Build GRCm38
Gene Family ID
MIPF0000056,
mir-148
Distance to Amplicon
171 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM12269
Ambion
®
Pre-miR
™
miRNA Precursor :
PM12269
TaqMan
™
MicroRNA Assay :
000475
mir
Vana
®
miRNA inhibitor :
MH12269
mir
Vana
®
miRNA mimic :
MC12269
TaqMan
™
Advanced miRNA Assay :
477921_mir
TaqMan
™
Advanced miRNA Assay :
mmu482952_mir
TaqMan
™
Advanced miRNA Assay :
rno481445_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM19559
Ambion
®
Pre-miR
™
miRNA Precursor :
PM19559
TaqMan
™
MicroRNA Assay :
464405_mat
mir
Vana
®
miRNA inhibitor :
MH19559
mir
Vana
®
miRNA mimic :
MC19559
TaqMan
™
Advanced miRNA Assay :
mmu481553_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline