Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM12387
          Assay Name: hsa-miR-10b-3p
          miRBase Accession Number: MI0000267
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 176150303 - 176150412 [+] on Build GRCh38
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Species: Human, Alligator mississippiensis, Cricetulus griseus, Eptesicus fuscus, Goat, Pteropus alecto
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM12387
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          ACAGAUUCGAUUCUAGGGGAAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-10b-3p MIMAT0004556
          Alligator mississippiensis ami-miR-10b-3p MIMAT0038033
          Cricetulus griseus cgr-miR-10b-3p MIMAT0023738
          Eptesicus fuscus efu-miR-10 MIMAT0034920
          Goat chi-miR-10b-3p MIMAT0035914
          Pteropus alecto pal-miR-10b-3p MIMAT0039900

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-10b
          Stem-loop Accession # MI0000267
          Stem-loop Sequence
          CCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCGUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA
          Chromosome Location Chr. 2 - 176150303 - 176150412 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004556
          miRBase ID: hsa-miR-10b-3p
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location: Chr. 2 - 176150303 - 176150412 [+] on Build GRCh38

          Alligator mississippiensis

          Stem-loop ID ami-mir-10b
          Stem-loop Accession # MI0029664
          Stem-loop Sequence
          CGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038033
          miRBase ID: ami-miR-10b-3p
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-10b
          Stem-loop Accession # MI0020382
          Stem-loop Sequence
          UGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUACCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023738
          miRBase ID: cgr-miR-10b-3p
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-10
          Stem-loop Accession # MI0028605
          Stem-loop Sequence
          GUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0034920
          miRBase ID: efu-miR-10
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-10b
          Stem-loop Accession # MI0030597
          Stem-loop Sequence
          CAGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCUCGUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035914
          miRBase ID: chi-miR-10b-3p
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-10b
          Stem-loop Accession # MI0032390
          Stem-loop Sequence
          UACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039900
          miRBase ID: pal-miR-10b-3p
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302879_pri
          Ambion® Pre-miR™ miRNA Precursor : PM12387
          TaqMan™ MicroRNA Assay : 002315
          mirVana® miRNA inhibitor : MH12387
          mirVana® miRNA mimic : MC12387
          TaqMan™ Advanced miRNA Assay : 477868_mir


          miRBase Alias:

          Human: hsa-miR-10b* (v17)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline