Search
Search

| Assay Name: | hsa-miR-10b-3p |
|---|---|
| miRBase Accession Number: | MI0000267 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACAGAUUCGAUUCUAGGGGAAU |
| Species: | Human, Alligator mississippiensis, Cricetulus griseus, Eptesicus fuscus, Goat, Pteropus alecto |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000033, mir-10 |
Mature miRNA Sequence: | ACAGAUUCGAUUCUAGGGGAAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-10b-3p | MIMAT0004556 |
| Alligator mississippiensis | ami-miR-10b-3p | MIMAT0038033 |
| Cricetulus griseus | cgr-miR-10b-3p | MIMAT0023738 |
| Eptesicus fuscus | efu-miR-10 | MIMAT0034920 |
| Goat | chi-miR-10b-3p | MIMAT0035914 |
| Pteropus alecto | pal-miR-10b-3p | MIMAT0039900 |