Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12387
          Assay Name: hsa-miR-10b-3p
          miRBase Accession Number: MI0000267
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 176150303 - 176150412 [+] on Build GRCh38
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Species: Human, Alligator mississippiensis, Cricetulus griseus, Eptesicus fuscus, Goat, Pteropus alecto
          Product Type: mirVana® miRNA mimic
          Assay ID MC12387

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          ACAGAUUCGAUUCUAGGGGAAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-10b-3p MIMAT0004556
          Alligator mississippiensis ami-miR-10b-3p MIMAT0038033
          Cricetulus griseus cgr-miR-10b-3p MIMAT0023738
          Eptesicus fuscus efu-miR-10 MIMAT0034920
          Goat chi-miR-10b-3p MIMAT0035914
          Pteropus alecto pal-miR-10b-3p MIMAT0039900

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-10b
          Stem-loop Accession # MI0000267
          Stem-loop Sequence
          CCAGAGGUUGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCGUAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCA
          Chromosome Location Chr. 2 - 176150303 - 176150412 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004556
          miRBase ID: hsa-miR-10b-3p
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location: Chr. 2 - 176150303 - 176150412 [+] on Build GRCh38

          Alligator mississippiensis

          Stem-loop ID ami-mir-10b
          Stem-loop Accession # MI0029664
          Stem-loop Sequence
          CGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUUCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038033
          miRBase ID: ami-miR-10b-3p
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-10b
          Stem-loop Accession # MI0020382
          Stem-loop Sequence
          UGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUACCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023738
          miRBase ID: cgr-miR-10b-3p
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-10
          Stem-loop Accession # MI0028605
          Stem-loop Sequence
          GUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0034920
          miRBase ID: efu-miR-10
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-10b
          Stem-loop Accession # MI0030597
          Stem-loop Sequence
          CAGUAACGUUGUCUAUAUAUACCCUGUAGAACCGAAUUUGUGUGGUAUCCUCGUAGUCACAGAUUCGAUUCUAGGGGAAUAUAUGGUCGAUGCAAAAACUUCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035914
          miRBase ID: chi-miR-10b-3p
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-10b
          Stem-loop Accession # MI0032390
          Stem-loop Sequence
          UACCCUGUAGAACCGAAUUUGUGUGGUAUCCACAUAGUCACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039900
          miRBase ID: pal-miR-10b-3p
          Mature miRNA Sequence: ACAGAUUCGAUUCUAGGGGAAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302879_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12387
          Ambion® Pre-miR™ miRNA Precursor : PM12387
          TaqMan™ MicroRNA Assay : 002315
          mirVana® miRNA inhibitor : MH12387
          TaqMan™ Advanced miRNA Assay : 477868_mir


          miRBase Alias:

          Human: hsa-miR-10b* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline