Search
Search

| Assay Name: | hsa-miR-518d-5p |
|---|---|
| miRBase Accession Number: | MI0003171 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUCUAGAGGGAAGCACUUUCUG |
| Species: | Human, Gorilla gorilla, Pan troglodytes, Pongo pygmaeus |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CUCUAGAGGGAAGCACUUUCUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-518d-5p | MIMAT0005456 |
| Human | hsa-miR-520c-5p | MIMAT0005455 |
| Human | hsa-miR-526a-5p | MIMAT0002845 |
| Gorilla gorilla | ggo-miR-518d-5p | MIMAT0036429 |
| Gorilla gorilla | ggo-miR-520c | MIMAT0036420 |
| Gorilla gorilla | ggo-miR-526a | MIMAT0036426 |
| View More Rows | ||
| Pan troglodytes | ptr-miR-526a | MIMAT0008210 |
| Pongo pygmaeus | ppy-miR-518e-5p | MIMAT0036450 |