Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM12843
          Assay Name: hsa-miR-518d-5p
          miRBase Accession Number: MI0003171
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 53734877 - 53734963 [+] on Build GRCh38
          Mature miRNA Sequence: CUCUAGAGGGAAGCACUUUCUG
          Species: Human, Gorilla gorilla, Pan troglodytes, Pongo pygmaeus
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM12843
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000020, mir-515

          Mature miRNA Sequence:

          CUCUAGAGGGAAGCACUUUCUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-518d-5p MIMAT0005456
          Human hsa-miR-520c-5p MIMAT0005455
          Human hsa-miR-526a-5p MIMAT0002845
          Gorilla gorilla ggo-miR-518d-5p MIMAT0036429
          Gorilla gorilla ggo-miR-520c MIMAT0036420
          Gorilla gorilla ggo-miR-526a MIMAT0036426
          View More Rows
          Pan troglodytes ptr-miR-526a MIMAT0008210
          Pongo pygmaeus ppy-miR-518e-5p MIMAT0036450

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-518d
          Stem-loop Accession # MI0003171
          Stem-loop Sequence
          UCCCAUGCUGUGACCCUCUAGAGGGAAGCACUUUCUGUUGUCUGAAAGAAACCAAAGCGCUUCCCUUUGGAGCGUUACGGUUUGAGA
          Chromosome Location Chr. 19 - 53734877 - 53734963 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005456
          miRBase ID: hsa-miR-518d-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCACUUUCUG
          Chromosome Location: Chr. 19 - 53734877 - 53734963 [+] on Build GRCh38

          Stem-loop ID hsa-mir-520c
          Stem-loop Accession # MI0003158
          Stem-loop Sequence
          UCUCAGGCUGUCGUCCUCUAGAGGGAAGCACUUUCUGUUGUCUGAAAGAAAAGAAAGUGCUUCCUUUUAGAGGGUUACCGUUUGAGA
          Chromosome Location Chr. 19 - 53707453 - 53707539 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005455
          miRBase ID: hsa-miR-520c-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCACUUUCUG
          Chromosome Location: Chr. 19 - 53707453 - 53707539 [+] on Build GRCh38

          Stem-loop ID hsa-mir-526a-1
          Stem-loop Accession # MI0003157
          Stem-loop Sequence
          CUCAGGCUGUGACCCUCUAGAGGGAAGCACUUUCUGUUGCUUGAAAGAAGAGAAAGCGCUUCCUUUUAGAGGAUUACUCUUUGAG
          Chromosome Location Chr. 19 - 53706252 - 53706336 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002845
          miRBase ID: hsa-miR-526a-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCACUUUCUG
          Chromosome Location: Chr. 19 - 53726922 - 53726986 [+] on Build GRCh38

          Stem-loop ID hsa-mir-526a-2
          Stem-loop Accession # MI0003168
          Stem-loop Sequence
          GUGACCCUCUAGAGGGAAGCACUUUCUGUUGAAAGAAAAGAACAUGCAUCCUUUCAGAGGGUUAC
          Chromosome Location Chr. 19 - 53726922 - 53726986 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002845
          miRBase ID: hsa-miR-526a-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCACUUUCUG
          Chromosome Location: Chr. 19 - 53726922 - 53726986 [+] on Build GRCh38

          Gorilla gorilla

          Stem-loop ID ggo-mir-518d
          Stem-loop Accession # MI0031083
          Stem-loop Sequence
          CUGGAGUGAGAAGAUCCCAUGCUGUGACCCUCUAGAGGGAAGCACUUUCUGUUGUCUGAAAGAAACCAAAGCGCUUCCCUUUGGAGCGUUACGGUUUGAGAAACUCAAUGUUGAAGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036429
          miRBase ID: ggo-miR-518d-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCACUUUCUG
          Chromosome Location: NA

          Stem-loop ID ggo-mir-520c
          Stem-loop Accession # MI0031077
          Stem-loop Sequence
          CAAGAAGAUCUUGGGCUGUCAUCCUCUAGAGGGAAGCACUUUCUGUUGUCUGAGAGAAAAGAAAGUGCUUCCUUUUAAAGGGUUACCGUUUGAGAAAAGCAACAUUGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036420
          miRBase ID: ggo-miR-520c
          Mature miRNA Sequence: CUCUAGAGGGAAGCACUUUCUG
          Chromosome Location: NA

          Stem-loop ID ggo-mir-526a
          Stem-loop Accession # MI0031081
          Stem-loop Sequence
          UAAGAAGAUCUCGUGCUGUGACCCUCUAGAGGGAAGCACUUUCUGUUGUCUGAAAGAAAAGAACAUGCAUCCUUUCAGAGGGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036426
          miRBase ID: ggo-miR-526a
          Mature miRNA Sequence: CUCUAGAGGGAAGCACUUUCUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-526a-1
          Stem-loop Accession # MI0008737
          Stem-loop Sequence
          UCAGGCUGUGACCCUCUAGAGGGAAGCACUUUCUGUUGCUUGAAAGAAGAGAAAGCGCUUCCUUUUAGAGGAUUACUCUUUGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008210
          miRBase ID: ptr-miR-526a
          Mature miRNA Sequence: CUCUAGAGGGAAGCACUUUCUG
          Chromosome Location: NA

          Stem-loop ID ptr-mir-526a-2
          Stem-loop Accession # MI0008738
          Stem-loop Sequence
          UGACCCUCUAGAGGGAAGCACUUUCUGUUGUCUGAAAGAAAAGAACAUGCAUCCUUUCAGAGGGUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008210
          miRBase ID: ptr-miR-526a
          Mature miRNA Sequence: CUCUAGAGGGAAGCACUUUCUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-518e
          Stem-loop Accession # MI0031104
          Stem-loop Sequence
          CUGGAGCAAGAAGAUCUCAUGAUGUGACCCCUCUAGAGGGAAGCACUUUCUGUUGGCUAAAAGAAAAAGAAAGCGCUUCCCUUCAGAGUGUUACGCUUUGAGAAAAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036450
          miRBase ID: ppy-miR-518e-5p
          Mature miRNA Sequence: CUCUAGAGGGAAGCACUUUCUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03293935_pri
          TaqMan™ Pri-miRNA Assay : Hs03293898_pri
          TaqMan™ Pri-miRNA Assay : Hs03304070_pri
          TaqMan™ Pri-miRNA Assay : Hs03304004_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12843
          TaqMan™ MicroRNA Assay : 002389
          mirVana® miRNA inhibitor : MH12843
          mirVana® miRNA mimic : MC12843
          TaqMan™ Advanced miRNA Assay : 479530_mir


          miRBase Alias:

          Human: hsa-miR-526a (v21)
          Pongo pygmaeus: ppy-miR-518g-5p (v21)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0