Search
Search

| Assay Name: | hsa-miR-518d-5p |
|---|---|
| miRBase Accession Number: | MI0003171 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUCUAGAGGGAAGCACUUUCUG |
| Species: | Human, Gorilla gorilla, Pan troglodytes, Pongo pygmaeus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CUCUAGAGGGAAGCACUUUCUG |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-518d-5p | MIMAT0005456 | |
| Human | hsa-miR-520c-5p | MIMAT0005455 | |
| Human | hsa-miR-526a-5p | MIMAT0002845 | |
| Gorilla gorilla | ggo-miR-518d-5p | MIMAT0036429 | |
| Gorilla gorilla | ggo-miR-520c | MIMAT0036420 | |
| Gorilla gorilla | ggo-miR-526a | MIMAT0036426 | |
| View More Rows | |||
| Pan troglodytes | ptr-miR-526a | MIMAT0008210 | |
| Pongo pygmaeus | ppy-miR-518e-5p | MIMAT0036450 | |