Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM12899
          Assay Name: hsa-miR-26b-5p
          miRBase Accession Number: MI0000084
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 218402646 - 218402722 [+] on Build GRCh38
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Species: Human, Mouse, Rat, Gorilla gorilla, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Sheep
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM12899
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000043, mir-26

          Mature miRNA Sequence:

          UUCAAGUAAUUCAGGAUAGGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-26b-5p MIMAT0000083
          Mouse mmu-miR-26b-5p MIMAT0000534
          Rat rno-miR-26b-5p MIMAT0000797
          Gorilla gorilla ggo-miR-26b MIMAT0024091
          Pan troglodytes ptr-miR-26b MIMAT0008077
          Pongo pygmaeus ppy-miR-26b MIMAT0015733
          View More Rows
          Rhesus monkey mml-miR-26b-5p MIMAT0006166
          Sheep oar-miR-26b MIMAT0030052

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-26b
          Stem-loop Accession # MI0000084
          Stem-loop Sequence
          CCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGUGCUGUCCAGCCUGUUCUCCAUUACUUGGCUCGGGGACCGG
          Chromosome Location Chr. 2 - 218402646 - 218402722 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000083
          miRBase ID: hsa-miR-26b-5p
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: Chr. 2 - 218402646 - 218402722 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-26b
          Stem-loop Accession # MI0000575
          Stem-loop Sequence
          UGCCCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGGUGCUGACCAGCCUGUUCUCCAUUACUUGGCUCGGGGGCCGGUGCC
          Chromosome Location Chr. 1 - 74394310 - 74394394 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000534
          miRBase ID: mmu-miR-26b-5p
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: Chr. 1 - 74394310 - 74394394 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-26b
          Stem-loop Accession # MI0000858
          Stem-loop Sequence
          UGCCCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGGUGCUGGCCAGCCUGUUCUCCAUUACUUGGCUCGGGGGCCGGUGCC
          Chromosome Location Chr. 9 - 81675275 - 81675359 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000797
          miRBase ID: rno-miR-26b-5p
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: Chr. 9 - 81675275 - 81675359 [+] on Build Rnor_6.0

          Gorilla gorilla

          Stem-loop ID ggo-mir-26b
          Stem-loop Accession # MI0020638
          Stem-loop Sequence
          CCACCCUGCCCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGUGCUGUCCAGCCUGUUCUCCAUUACUUGGCUCGGGGACCGGUGCCCUGCAGCCUUGGGGUGAGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024091
          miRBase ID: ggo-miR-26b
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-26b
          Stem-loop Accession # MI0008590
          Stem-loop Sequence
          CGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGUGCUGUCCAGCCUGUUCUCCAUUACUUGGCUCGGGGACCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008077
          miRBase ID: ptr-miR-26b
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-26b
          Stem-loop Accession # MI0014799
          Stem-loop Sequence
          CCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGUGCUGUCCAGCCUGUUCUCCAUUACUUGGCUCGGGGACCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015733
          miRBase ID: ppy-miR-26b
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-26b
          Stem-loop Accession # MI0007594
          Stem-loop Sequence
          CCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGUGCUGUCCAGCCUGUUCUCCAUUACUUGGCUCGGGGACCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006166
          miRBase ID: mml-miR-26b-5p
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-26b
          Stem-loop Accession # MI0025274
          Stem-loop Sequence
          UGCCCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGUGCUGUCCAGCCUGUUCUCCAUUACUUGGCUCGGGGGCCGGUGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030052
          miRBase ID: oar-miR-26b
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302654_pri
          TaqMan™ Pri-miRNA Assay : Rn03465237_pri
          TaqMan™ Pri-miRNA Assay : Mm03306849_pri
          Ambion® Pre-miR™ miRNA Precursor : PM12899
          TaqMan™ MicroRNA Assay : 000407
          mirVana® miRNA inhibitor : MH12899
          mirVana® miRNA mimic : MC12899
          TaqMan™ Advanced miRNA Assay : 478418_mir
          TaqMan™ Advanced miRNA Assay : rno481461_mir
          TaqMan™ Advanced miRNA Assay : mmu482965_mir


          miRBase Alias:

          Human: hsa-miR-26b (v17)
          Mouse: mmu-miR-26b (v17)
          Rat: rno-miR-26b (v18)
          Macaca mulatta: mml-miR-26b (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline