Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478418_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-26b-5p
          Stem-loop Accession Number: MI0000084
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 218402646 - 218402722 [+] on Build GRCh38
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Species: Human, Mouse, Rat, Gorilla gorilla, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Sheep
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478418_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000043, mir-26

          Mature miRNA Sequence:

          UUCAAGUAAUUCAGGAUAGGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-26b-5p MIMAT0000083
          Mouse mmu-miR-26b-5p MIMAT0000534
          Rat rno-miR-26b-5p MIMAT0000797
          Gorilla gorilla ggo-miR-26b MIMAT0024091
          Pan troglodytes ptr-miR-26b MIMAT0008077
          Pongo pygmaeus ppy-miR-26b MIMAT0015733
          View More Rows
          Rhesus monkey mml-miR-26b-5p MIMAT0006166
          Sheep oar-miR-26b MIMAT0030052

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-26b
          Stem-loop Accession # MI0000084
          Stem-loop Sequence
          CCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGUGCUGUCCAGCCUGUUCUCCAUUACUUGGCUCGGGGACCGG
          Chromosome Location Chr. 2 - 218402646 - 218402722 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000083
          miRBase ID: hsa-miR-26b-5p
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: Chr. 2 - 218402646 - 218402722 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-26b
          Stem-loop Accession # MI0000575
          Stem-loop Sequence
          UGCCCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGGUGCUGACCAGCCUGUUCUCCAUUACUUGGCUCGGGGGCCGGUGCC
          Chromosome Location Chr. 1 - 74394310 - 74394394 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000534
          miRBase ID: mmu-miR-26b-5p
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: Chr. 1 - 74394310 - 74394394 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-26b
          Stem-loop Accession # MI0000858
          Stem-loop Sequence
          UGCCCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGGUGCUGGCCAGCCUGUUCUCCAUUACUUGGCUCGGGGGCCGGUGCC
          Chromosome Location Chr. 9 - 81675275 - 81675359 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000797
          miRBase ID: rno-miR-26b-5p
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: Chr. 9 - 81675275 - 81675359 [+] on Build Rnor_6.0

          Gorilla gorilla

          Stem-loop ID ggo-mir-26b
          Stem-loop Accession # MI0020638
          Stem-loop Sequence
          CCACCCUGCCCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGUGCUGUCCAGCCUGUUCUCCAUUACUUGGCUCGGGGACCGGUGCCCUGCAGCCUUGGGGUGAGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024091
          miRBase ID: ggo-miR-26b
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-26b
          Stem-loop Accession # MI0008590
          Stem-loop Sequence
          CGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGUGCUGUCCAGCCUGUUCUCCAUUACUUGGCUCGGGGACCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008077
          miRBase ID: ptr-miR-26b
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-26b
          Stem-loop Accession # MI0014799
          Stem-loop Sequence
          CCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGUGCUGUCCAGCCUGUUCUCCAUUACUUGGCUCGGGGACCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015733
          miRBase ID: ppy-miR-26b
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-26b
          Stem-loop Accession # MI0007594
          Stem-loop Sequence
          CCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGUGCUGUCCAGCCUGUUCUCCAUUACUUGGCUCGGGGACCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006166
          miRBase ID: mml-miR-26b-5p
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-26b
          Stem-loop Accession # MI0025274
          Stem-loop Sequence
          UGCCCGGGACCCAGUUCAAGUAAUUCAGGAUAGGUUGUGUGCUGUCCAGCCUGUUCUCCAUUACUUGGCUCGGGGGCCGGUGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030052
          miRBase ID: oar-miR-26b
          Mature miRNA Sequence: UUCAAGUAAUUCAGGAUAGGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302654_pri
          TaqMan™ Pri-miRNA Assay : Rn03465237_pri
          TaqMan™ Pri-miRNA Assay : Mm03306849_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12899
          Ambion® Pre-miR™ miRNA Precursor : PM12899
          TaqMan™ MicroRNA Assay : 000407
          mirVana® miRNA inhibitor : MH12899
          mirVana® miRNA mimic : MC12899
          TaqMan™ Advanced miRNA Assay : rno481461_mir
          TaqMan™ Advanced miRNA Assay : mmu482965_mir
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human A Card

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline