Search
Search

| Assay Name: | hsa-miR-26b-5p |
|---|---|
| miRBase Accession Number: | MI0000084 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUCAAGUAAUUCAGGAUAGGU |
| Species: | Human, Mouse, Rat, Gorilla gorilla, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Sheep |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000043, mir-26 |
Mature miRNA Sequence: | UUCAAGUAAUUCAGGAUAGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-26b-5p | MIMAT0000083 |
| Mouse | mmu-miR-26b-5p | MIMAT0000534 |
| Rat | rno-miR-26b-5p | MIMAT0000797 |
| Gorilla gorilla | ggo-miR-26b | MIMAT0024091 |
| Pan troglodytes | ptr-miR-26b | MIMAT0008077 |
| Pongo pygmaeus | ppy-miR-26b | MIMAT0015733 |
| View More Rows | ||
| Rhesus monkey | mml-miR-26b-5p | MIMAT0006166 |
| Sheep | oar-miR-26b | MIMAT0030052 |