Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM12933
          Assay Name: hsa-miR-28-3p
          miRBase Accession Number: MI0000086
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 188688781 - 188688866 [+] on Build GRCh38
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Species: Human, Rat, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Horse, Microcebus murinus, Nomascus leucogenys, Pig, Pteropus alecto, Rhesus monkey
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM12933
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000057, mir-28

          Mature miRNA Sequence:

          CACUAGAUUGUGAGCUCCUGGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-28-3p MIMAT0004502
          Rat rno-miR-28-3p MIMAT0004716
          Cricetulus griseus cgr-miR-28-3p MIMAT0023879
          Dasypus novemcinctus dno-miR-28-3p MIMAT0047548
          Daubentonia madagascariensis dma-miR-28 MIMAT0049257
          Dog cfa-miR-28 MIMAT0006675
          View More Rows
          Horse eca-miR-28-3p MIMAT0013093
          Microcebus murinus mmr-miR-28 MIMAT0049146
          Nomascus leucogenys nle-miR-28 MIMAT0049459
          Pig ssc-miR-28-3p MIMAT0015211
          Pteropus alecto pal-miR-28-3p MIMAT0039891
          Rhesus monkey mml-miR-28-3p MIMAT0026575

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-28
          Stem-loop Accession # MI0000086
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location Chr. 3 - 188688781 - 188688866 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004502
          miRBase ID: hsa-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: Chr. 3 - 188688781 - 188688866 [+] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-28
          Stem-loop Accession # MI0000861
          Stem-loop Sequence
          GGUCCCUACCCGCAAGGAGCUCACAGUCUAUUGAGUUCCUUUUCUGAUUCUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location Chr. 11 - 79380523 - 79380608 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004716
          miRBase ID: rno-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: Chr. 11 - 79380523 - 79380608 [-] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-28
          Stem-loop Accession # MI0020475
          Stem-loop Sequence
          CCCUACCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGAUUCUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023879
          miRBase ID: cgr-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-28
          Stem-loop Accession # MI0038929
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUUCCUUUUUGACUUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047548
          miRBase ID: dno-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-28
          Stem-loop Accession # MI0039985
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUUCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049257
          miRBase ID: dma-miR-28
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-28
          Stem-loop Accession # MI0008082
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006675
          miRBase ID: cfa-miR-28
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-28
          Stem-loop Accession # MI0012840
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013093
          miRBase ID: eca-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-28
          Stem-loop Accession # MI0039860
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049146
          miRBase ID: mmr-miR-28
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-28
          Stem-loop Accession # MI0040212
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGACCUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049459
          miRBase ID: nle-miR-28
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-28
          Stem-loop Accession # MI0002430
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGUCUUCCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015211
          miRBase ID: ssc-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-28
          Stem-loop Accession # MI0032385
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUGCUUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039891
          miRBase ID: pal-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-28
          Stem-loop Accession # MI0002649
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUUCCUUUGUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026575
          miRBase ID: mml-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465014_pri
          TaqMan™ Pri-miRNA Assay : Hs03302668_pri
          Ambion® Pre-miR™ miRNA Precursor : PM12933
          TaqMan™ MicroRNA Assay : 002446
          mirVana® miRNA inhibitor : MH12933
          mirVana® miRNA mimic : MC12933
          TaqMan™ Advanced miRNA Assay : rno481017_mir
          TaqMan™ Advanced miRNA Assay : 477999_mir


          miRBase Alias:

          Rat: rno-miR-28* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline