Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12933
          Assay Name: hsa-miR-28-3p
          miRBase Accession Number: MI0000086
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 188688781 - 188688866 [+] on Build GRCh38
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Species: Human, Rat, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Horse, Microcebus murinus, Nomascus leucogenys, Pig, Pteropus alecto, Rhesus monkey
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12933

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000057, mir-28

          Mature miRNA Sequence:

          CACUAGAUUGUGAGCUCCUGGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-28-3p MIMAT0004502
          Rat rno-miR-28-3p MIMAT0004716
          Cricetulus griseus cgr-miR-28-3p MIMAT0023879
          Dasypus novemcinctus dno-miR-28-3p MIMAT0047548
          Daubentonia madagascariensis dma-miR-28 MIMAT0049257
          Dog cfa-miR-28 MIMAT0006675
          View More Rows
          Horse eca-miR-28-3p MIMAT0013093
          Microcebus murinus mmr-miR-28 MIMAT0049146
          Nomascus leucogenys nle-miR-28 MIMAT0049459
          Pig ssc-miR-28-3p MIMAT0015211
          Pteropus alecto pal-miR-28-3p MIMAT0039891
          Rhesus monkey mml-miR-28-3p MIMAT0026575

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-28
          Stem-loop Accession # MI0000086
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location Chr. 3 - 188688781 - 188688866 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004502
          miRBase ID: hsa-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: Chr. 3 - 188688781 - 188688866 [+] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-28
          Stem-loop Accession # MI0000861
          Stem-loop Sequence
          GGUCCCUACCCGCAAGGAGCUCACAGUCUAUUGAGUUCCUUUUCUGAUUCUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location Chr. 11 - 79380523 - 79380608 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004716
          miRBase ID: rno-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: Chr. 11 - 79380523 - 79380608 [-] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-28
          Stem-loop Accession # MI0020475
          Stem-loop Sequence
          CCCUACCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGAUUCUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023879
          miRBase ID: cgr-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-28
          Stem-loop Accession # MI0038929
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUUCCUUUUUGACUUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047548
          miRBase ID: dno-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-28
          Stem-loop Accession # MI0039985
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUUCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049257
          miRBase ID: dma-miR-28
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-28
          Stem-loop Accession # MI0008082
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006675
          miRBase ID: cfa-miR-28
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-28
          Stem-loop Accession # MI0012840
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013093
          miRBase ID: eca-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-28
          Stem-loop Accession # MI0039860
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049146
          miRBase ID: mmr-miR-28
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-28
          Stem-loop Accession # MI0040212
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGACCUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049459
          miRBase ID: nle-miR-28
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-28
          Stem-loop Accession # MI0002430
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGUCUUCCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015211
          miRBase ID: ssc-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-28
          Stem-loop Accession # MI0032385
          Stem-loop Sequence
          AAGGAGCUCACAGUCUAUUGAGUUGCUUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039891
          miRBase ID: pal-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-28
          Stem-loop Accession # MI0002649
          Stem-loop Sequence
          GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUUCCUUUGUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026575
          miRBase ID: mml-miR-28-3p
          Mature miRNA Sequence: CACUAGAUUGUGAGCUCCUGGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465014_pri
          TaqMan™ Pri-miRNA Assay : Hs03302668_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12933
          Ambion® Pre-miR™ miRNA Precursor : PM12933
          TaqMan™ MicroRNA Assay : 002446
          mirVana® miRNA mimic : MC12933
          TaqMan™ Advanced miRNA Assay : rno481017_mir
          TaqMan™ Advanced miRNA Assay : 477999_mir


          miRBase Alias:

          Rat: rno-miR-28* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline