Search
Search

| Assay Name: | hsa-miR-28-3p |
|---|---|
| miRBase Accession Number: | MI0000086 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CACUAGAUUGUGAGCUCCUGGA |
| Species: | Human, Rat, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Horse, Microcebus murinus, Nomascus leucogenys, Pig, Pteropus alecto, Rhesus monkey |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | CACUAGAUUGUGAGCUCCUGGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-28-3p | MIMAT0004502 |
| Rat | rno-miR-28-3p | MIMAT0004716 |
| Cricetulus griseus | cgr-miR-28-3p | MIMAT0023879 |
| Dasypus novemcinctus | dno-miR-28-3p | MIMAT0047548 |
| Daubentonia madagascariensis | dma-miR-28 | MIMAT0049257 |
| Dog | cfa-miR-28 | MIMAT0006675 |
| View More Rows | ||
| Horse | eca-miR-28-3p | MIMAT0013093 |
| Microcebus murinus | mmr-miR-28 | MIMAT0049146 |
| Nomascus leucogenys | nle-miR-28 | MIMAT0049459 |
| Pig | ssc-miR-28-3p | MIMAT0015211 |
| Pteropus alecto | pal-miR-28-3p | MIMAT0039891 |
| Rhesus monkey | mml-miR-28-3p | MIMAT0026575 |