Search
Search

| Assay Name: | hsa-let-7g-3p |
|---|---|
| miRBase Accession Number: | MI0000433 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGUACAGGCCACUGCCUUGC |
| Species: | Human, Rat, Alligator mississippiensis, Chrysemys picta, Cricetulus griseus, Python bivittatus, Taeniopygia guttata |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | CUGUACAGGCCACUGCCUUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-let-7g-3p | MIMAT0004584 |
| Rat | rno-let-7g-3p | MIMAT0035720 |
| Alligator mississippiensis | ami-let-7g-3p | MIMAT0038015 |
| Chrysemys picta | cpi-let-7g-3p | MIMAT0037609 |
| Cricetulus griseus | cgr-let-7g-3p | MIMAT0023723 |
| Python bivittatus | pbv-let-7g-3p | MIMAT0038809 |
| View More Rows | ||
| Taeniopygia guttata | tgu-let-7g-3p | MIMAT0026986 |