Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 477850_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-let-7g-3p
          Stem-loop Accession Number: MI0000433
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 52268278 - 52268361 [-] on Build GRCh38
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Species: Human, Rat, Alligator mississippiensis, Chrysemys picta, Cricetulus griseus, Python bivittatus, Taeniopygia guttata
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 477850_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          CUGUACAGGCCACUGCCUUGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7g-3p MIMAT0004584
          Rat rno-let-7g-3p MIMAT0035720
          Alligator mississippiensis ami-let-7g-3p MIMAT0038015
          Chrysemys picta cpi-let-7g-3p MIMAT0037609
          Cricetulus griseus cgr-let-7g-3p MIMAT0023723
          Python bivittatus pbv-let-7g-3p MIMAT0038809
          View More Rows
          Taeniopygia guttata tgu-let-7g-3p MIMAT0026986

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7g
          Stem-loop Accession # MI0000433
          Stem-loop Sequence
          AGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCA
          Chromosome Location Chr. 3 - 52268278 - 52268361 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004584
          miRBase ID: hsa-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: Chr. 3 - 52268278 - 52268361 [-] on Build GRCh38

          Rat

          Stem-loop ID rno-let-7g
          Stem-loop Accession # MI0030347
          Stem-loop Sequence
          CCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGG
          Chromosome Location Chr. 8 - 114874457 - 114874544 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0035720
          miRBase ID: rno-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: Chr. 8 - 114874457 - 114874544 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-let-7g
          Stem-loop Accession # MI0029650
          Stem-loop Sequence
          GCCUGAUUCCAAGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUUAACUGUACAGGCCACUGCCUUGCCUGGGACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038015
          miRBase ID: ami-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-let-7g
          Stem-loop Accession # MI0029382
          Stem-loop Sequence
          UCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037609
          miRBase ID: cpi-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-let-7g
          Stem-loop Accession # MI0020373
          Stem-loop Sequence
          AUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGGAACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023723
          miRBase ID: cgr-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-let-7g
          Stem-loop Accession # MI0030141
          Stem-loop Sequence
          GUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUACGAUACCGCCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038809
          miRBase ID: pbv-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-let-7g-1
          Stem-loop Accession # MI0013740
          Stem-loop Sequence
          GAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026986
          miRBase ID: tgu-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: NA

          Stem-loop ID tgu-let-7g-2
          Stem-loop Accession # MI0013741
          Stem-loop Sequence
          GAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026986
          miRBase ID: tgu-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303029_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12996
          Ambion® Pre-miR™ miRNA Precursor : PM12996
          TaqMan™ MicroRNA Assay : 002118
          mirVana® miRNA inhibitor : MH12996
          mirVana® miRNA mimic : MC12996
          TaqMan™ Advanced miRNA Assay : rno481446_mir
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human B Card
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline