Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002118
          Assay Name: hsa-let-7g*
          miRBase Accession Number: MI0000433
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 52268278 - 52268361 [-] on Build GRCh38
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Species: Human, Rat, Alligator mississippiensis, Chrysemys picta, Cricetulus griseus, Python bivittatus, Taeniopygia guttata
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002118
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          CUGUACAGGCCACUGCCUUGC

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7g-3p MIMAT0004584
          Rat rno-let-7g-3p MIMAT0035720
          Alligator mississippiensis ami-let-7g-3p MIMAT0038015
          Chrysemys picta cpi-let-7g-3p MIMAT0037609
          Cricetulus griseus cgr-let-7g-3p MIMAT0023723
          Python bivittatus pbv-let-7g-3p MIMAT0038809
          View More Rows
          Taeniopygia guttata tgu-let-7g-3p MIMAT0026986

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7g
          Stem-loop Accession # MI0000433
          Stem-loop Sequence
          AGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCA
          Chromosome Location Chr. 3 - 52268278 - 52268361 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004584
          miRBase ID: hsa-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: Chr. 3 - 52268278 - 52268361 [-] on Build GRCh38

          Rat

          Stem-loop ID rno-let-7g
          Stem-loop Accession # MI0030347
          Stem-loop Sequence
          CCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGG
          Chromosome Location Chr. 8 - 114874457 - 114874544 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0035720
          miRBase ID: rno-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: Chr. 8 - 114874457 - 114874544 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-let-7g
          Stem-loop Accession # MI0029650
          Stem-loop Sequence
          GCCUGAUUCCAAGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUUAACUGUACAGGCCACUGCCUUGCCUGGGACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038015
          miRBase ID: ami-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-let-7g
          Stem-loop Accession # MI0029382
          Stem-loop Sequence
          UCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037609
          miRBase ID: cpi-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-let-7g
          Stem-loop Accession # MI0020373
          Stem-loop Sequence
          AUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCAGGAACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023723
          miRBase ID: cgr-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-let-7g
          Stem-loop Accession # MI0030141
          Stem-loop Sequence
          GUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUACGAUACCGCCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038809
          miRBase ID: pbv-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-let-7g-1
          Stem-loop Accession # MI0013740
          Stem-loop Sequence
          GAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026986
          miRBase ID: tgu-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: NA

          Stem-loop ID tgu-let-7g-2
          Stem-loop Accession # MI0013741
          Stem-loop Sequence
          GAUUCCAGGCUGAGGUAGUAGUUUGUACAGUUUGAGGGUCUAUGAUACCACCCGGUACAGGAGAUAACUGUACAGGCCACUGCCUUGCCUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026986
          miRBase ID: tgu-let-7g-3p
          Mature miRNA Sequence: CUGUACAGGCCACUGCCUUGC
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Human Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Human MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303029_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12996
          Ambion® Pre-miR™ miRNA Precursor : PM12996
          mirVana® miRNA inhibitor : MH12996
          mirVana® miRNA mimic : MC12996
          TaqMan™ Advanced miRNA Assay : 477850_mir
          TaqMan™ Advanced miRNA Assay : rno481446_mir
          miRNA Card : TaqMan® Human MicroRNA B Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          FGMIR : TaqManTM Human MicroRNA Array B v3
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0


          miRBase Alias:

          Human: hsa-let-7g* (v17)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline