Search
Search

| Assay Name: | rno-miR-33-3p |
|---|---|
| miRBase Accession Number: | MI0000874 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAUGUUUCCACAGUGCAUCA |
| Species: | Rat, Goat, Guinea pig |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000070, mir-33 |
Mature miRNA Sequence: | CAAUGUUUCCACAGUGCAUCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-33-3p | MIMAT0017104 |
| Goat | chi-miR-33a-3p | MIMAT0036152 |
| Guinea pig | cpo-miR-33a-3p | MIMAT0046919 |