Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481061_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-33-3p
          Stem-loop Accession Number: MI0000874
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 123431612 - 123431680 [+] on Build Rnor_6.0
          Mature miRNA Sequence: CAAUGUUUCCACAGUGCAUCA
          Species: Rat, Goat, Guinea pig
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481061_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000070, mir-33

          Mature miRNA Sequence:

          CAAUGUUUCCACAGUGCAUCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-33-3p MIMAT0017104
          Goat chi-miR-33a-3p MIMAT0036152
          Guinea pig cpo-miR-33a-3p MIMAT0046919

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-33
          Stem-loop Accession # MI0000874
          Stem-loop Sequence
          CCGUGGUGCAUUGUAGUUGCAUUGCAUGUUCUGGCAGUACCUGUGCAAUGUUUCCACAGUGCAUCACGG
          Chromosome Location Chr. 7 - 123431612 - 123431680 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017104
          miRBase ID: rno-miR-33-3p
          Mature miRNA Sequence: CAAUGUUUCCACAGUGCAUCA
          Chromosome Location: Chr. 7 - 123431612 - 123431680 [+] on Build Rnor_6.0

          Goat

          Stem-loop ID chi-mir-33a
          Stem-loop Accession # MI0030749
          Stem-loop Sequence
          UGCUGCGGUGCAUUGUAGUUGCAUUGCAUGUUCCGGCGGCACCGUGCAAUGUUUCCACAGUGCAUCACAGAGGCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036152
          miRBase ID: chi-miR-33a-3p
          Mature miRNA Sequence: CAAUGUUUCCACAGUGCAUCA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-33a
          Stem-loop Accession # MI0038598
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGUUCUGGCAGUACCGUGCAAUGUUUCCACAGUGCAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046919
          miRBase ID: cpo-miR-33a-3p
          Mature miRNA Sequence: CAAUGUUUCCACAGUGCAUCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465263_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM20107
          Ambion® Pre-miR™ miRNA Precursor : PM20107
          TaqMan™ MicroRNA Assay : 461759_mat
          mirVana® miRNA inhibitor : MH20107
          mirVana® miRNA mimic : MC20107

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline