Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03465263_pri
          Assay Name:
          rno-mir-33
          Stem-loop ID:
          rno-mir-33
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.7: 123431612 - 123431680 [+] on Build Rnor_6.0
          Stem-loop Location:
          Chr.7: 123431612 - 123431680 [+] on Build Rnor_6.0
          Mature miRNA ID:
          rno-miR-33-3p rno-miR-33-5p
          Species:
          Rat
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Rn03465263_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          82

          Chromosome Location:

          Chr. 7 - 123431462 - 123431462 [+] on 123431462 Build Rnor_6.0

          Species:

          Rat

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-33-3p MIMAT0017104
          Rat rno-miR-33-5p MIMAT0000812

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-33
          Stem-loop Accession # MI0000874
          Stem-loop Sequence
          CCGUGGUGCAUUGUAGUUGCAUUGCAUGUUCUGGCAGUACCUGUGCAAUGUUUCCACAGUGCAUCACGG
          Chromosome Location Chr. 7 - 123431612 - 123431680 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017104
          miRBase ID: rno-miR-33-3p
          Mature miRNA Sequence: CAAUGUUUCCACAGUGCAUCA
          Chromosome Location: Chr. 7 - 123431612 - 123431680 [+] on Build Rnor_6.0

          miRBase Accession #: MIMAT0000812
          miRBase ID: rno-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: Chr. 7 - 123431612 - 123431680 [+] on Build Rnor_6.0

          Gene Family ID MIPF0000070, mir-33
          Distance to Amplicon 116 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM20107
          Ambion® Pre-miR™ miRNA Precursor : PM20107
          TaqMan™ MicroRNA Assay : 461759_mat
          mirVana® miRNA inhibitor : MH20107
          mirVana® miRNA mimic : MC20107
          TaqMan™ Advanced miRNA Assay : rno481061_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12410
          Ambion® Pre-miR™ miRNA Precursor : PM12410
          TaqMan™ MicroRNA Assay : 465396_mat
          TaqMan™ MicroRNA Assay : 002135
          mirVana® miRNA inhibitor : MH12410
          mirVana® miRNA mimic : MC12410
          TaqMan™ Advanced miRNA Assay : rno481363_mir
          TaqMan™ Advanced miRNA Assay : 478347_mir
          TaqMan™ Advanced miRNA Assay : mmu482743_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline