Search
Search

| Assay Name: | rno-miR-33-5p |
|---|---|
| Stem-loop Accession Number: | MI0000091 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUGCAUUGUAGUUGCAUUGCA |
| Species: | Human, Mouse, Rat, Aedes aegypti, Alligator mississippiensis, Anolis carolinensis, Anopheles gambiae, Bombyx mori, Bovine, Branchiostoma belcheri, Capitella teleta, Chrysemys picta, Columba livia, Culex quinquefasciatus, Dasypus novemcinctus, Dinoponera quadriceps, Gadus morhua, Guinea pig, Horse, Manduca sexta, Monodelphis domestica, Oreochromis niloticus, Pteropus alecto, Python bivittatus, Rabbit, Salmo salar, Triops cancriformis, Tupaia chinensis, Xenopus laevis |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000070, mir-33 |
Mature miRNA Sequence: | GUGCAUUGUAGUUGCAUUGCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-33a-5p | MIMAT0000091 |
| Mouse | mmu-miR-33-5p | MIMAT0000667 |
| Rat | rno-miR-33-5p | MIMAT0000812 |
| Aedes aegypti | aae-miR-33 | MIMAT0014287 |
| Alligator mississippiensis | ami-miR-33-5p | MIMAT0038092 |
| Anolis carolinensis | aca-miR-33-5p | MIMAT0021934 |
| View More Rows | ||
| Anopheles gambiae | aga-miR-33-5p | MIMAT0041523 |
| Bombyx mori | bmo-miR-33-5p | MIMAT0013599 |
| Bovine | bta-miR-33a | MIMAT0009294 |
| Branchiostoma belcheri | bbe-miR-33-5p | MIMAT0031668 |
| Capitella teleta | cte-miR-33 | MIMAT0009553 |
| Chrysemys picta | cpi-miR-33-5p | MIMAT0037689 |
| Columba livia | cli-miR-33-5p | MIMAT0038464 |
| Culex quinquefasciatus | cqu-miR-33 | MIMAT0014431 |
| Dasypus novemcinctus | dno-miR-33a-5p | MIMAT0047571 |
| Dinoponera quadriceps | dqu-miR-33-5p | MIMAT0045665 |
| Gadus morhua | gmo-miR-33b-5p | MIMAT0044147 |
| Guinea pig | cpo-miR-33a-5p | MIMAT0046918 |
| Horse | eca-miR-33a | MIMAT0013189 |
| Manduca sexta | mse-miR-33 | MIMAT0024429 |
| Monodelphis domestica | mdo-miR-33-5p | MIMAT0012754 |
| Oreochromis niloticus | oni-miR-33a-5p | MIMAT0042819 |
| Oreochromis niloticus | oni-miR-33b-5p | MIMAT0042753 |
| Pteropus alecto | pal-miR-33a-5p | MIMAT0040081 |
| Python bivittatus | pbv-miR-33-5p | MIMAT0038882 |
| Rabbit | ocu-miR-33a-5p | MIMAT0048165 |
| Salmo salar | ssa-miR-33a-5p | MIMAT0032618 |
| Salmo salar | ssa-miR-33b-5p | MIMAT0032620 |
| Triops cancriformis | tcf-miR-33 | MIMAT0041358 |
| Tupaia chinensis | tch-miR-33a-5p | MIMAT0036468 |
| Xenopus laevis | xla-miR-33a-5p | MIMAT0046580 |