Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM12410
          Assay Name: hsa-miR-33a-5p
          miRBase Accession Number: MI0000091
          miRBase Version: v22.1
          Chromosome Location: Chr.22: 41900944 - 41901012 [+] on Build GRCh38
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Species: Human, Mouse, Rat, Aedes aegypti, Alligator mississippiensis, Anolis carolinensis, Anopheles gambiae, Bombyx mori, Bovine, Branchiostoma belcheri, Capitella teleta, Chrysemys picta, Columba livia, Culex quinquefasciatus, Dasypus novemcinctus, Dinoponera quadriceps, Gadus morhua, Guinea pig, Horse, Manduca sexta, Monodelphis domestica, Oreochromis niloticus, Pteropus alecto, Python bivittatus, Rabbit, Salmo salar, Triops cancriformis, Tupaia chinensis, Xenopus laevis
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM12410
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000070, mir-33

          Mature miRNA Sequence:

          GUGCAUUGUAGUUGCAUUGCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-33a-5p MIMAT0000091
          Mouse mmu-miR-33-5p MIMAT0000667
          Rat rno-miR-33-5p MIMAT0000812
          Aedes aegypti aae-miR-33 MIMAT0014287
          Alligator mississippiensis ami-miR-33-5p MIMAT0038092
          Anolis carolinensis aca-miR-33-5p MIMAT0021934
          View More Rows
          Anopheles gambiae aga-miR-33-5p MIMAT0041523
          Bombyx mori bmo-miR-33-5p MIMAT0013599
          Bovine bta-miR-33a MIMAT0009294
          Branchiostoma belcheri bbe-miR-33-5p MIMAT0031668
          Capitella teleta cte-miR-33 MIMAT0009553
          Chrysemys picta cpi-miR-33-5p MIMAT0037689
          Columba livia cli-miR-33-5p MIMAT0038464
          Culex quinquefasciatus cqu-miR-33 MIMAT0014431
          Dasypus novemcinctus dno-miR-33a-5p MIMAT0047571
          Dinoponera quadriceps dqu-miR-33-5p MIMAT0045665
          Gadus morhua gmo-miR-33b-5p MIMAT0044147
          Guinea pig cpo-miR-33a-5p MIMAT0046918
          Horse eca-miR-33a MIMAT0013189
          Manduca sexta mse-miR-33 MIMAT0024429
          Monodelphis domestica mdo-miR-33-5p MIMAT0012754
          Oreochromis niloticus oni-miR-33a-5p MIMAT0042819
          Oreochromis niloticus oni-miR-33b-5p MIMAT0042753
          Pteropus alecto pal-miR-33a-5p MIMAT0040081
          Python bivittatus pbv-miR-33-5p MIMAT0038882
          Rabbit ocu-miR-33a-5p MIMAT0048165
          Salmo salar ssa-miR-33a-5p MIMAT0032618
          Salmo salar ssa-miR-33b-5p MIMAT0032620
          Triops cancriformis tcf-miR-33 MIMAT0041358
          Tupaia chinensis tch-miR-33a-5p MIMAT0036468
          Xenopus laevis xla-miR-33a-5p MIMAT0046580

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-33a
          Stem-loop Accession # MI0000091
          Stem-loop Sequence
          CUGUGGUGCAUUGUAGUUGCAUUGCAUGUUCUGGUGGUACCCAUGCAAUGUUUCCACAGUGCAUCACAG
          Chromosome Location Chr. 22 - 41900944 - 41901012 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000091
          miRBase ID: hsa-miR-33a-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: Chr. 22 - 41900944 - 41901012 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-33
          Stem-loop Accession # MI0000707
          Stem-loop Sequence
          CUGUGGUGCAUUGUAGUUGCAUUGCAUGUUCUGGCAAUACCUGUGCAAUGUUUCCACAGUGCAUCACGG
          Chromosome Location Chr. 15 - 82198122 - 82198190 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000667
          miRBase ID: mmu-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: Chr. 15 - 82198122 - 82198190 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-33
          Stem-loop Accession # MI0000874
          Stem-loop Sequence
          CCGUGGUGCAUUGUAGUUGCAUUGCAUGUUCUGGCAGUACCUGUGCAAUGUUUCCACAGUGCAUCACGG
          Chromosome Location Chr. 7 - 123431612 - 123431680 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000812
          miRBase ID: rno-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: Chr. 7 - 123431612 - 123431680 [+] on Build Rnor_6.0

          Aedes aegypti

          Stem-loop ID aae-mir-33
          Stem-loop Accession # MI0013493
          Stem-loop Sequence
          CCACCCGGGUGCAUUGUAGUUGCAUUGCACUUUUGCGUUCGAUAACACGGUGCAAUACUUCUGCAAUGCAAGCCAGGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014287
          miRBase ID: aae-miR-33
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Alligator mississippiensis

          Stem-loop ID ami-mir-33-1
          Stem-loop Accession # MI0029698
          Stem-loop Sequence
          GGGUGACCGCUGUAGUGCAUUGUAGUUGCAUUGCAUGUUCUGGUAGUAUCUGUGCAAUGUUUCUGCAGUGCAGUAUAGAGGCACUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038092
          miRBase ID: ami-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Stem-loop ID ami-mir-33-2
          Stem-loop Accession # MI0029699
          Stem-loop Sequence
          GCAGUGGCCUCAGCUGUGGUGCAUUGUAGUUGCAUUGCAUGUGUCACAGAGGUGUGCAAUGCCUCUGCAGUGCAGCCCAGAGGAGGCUCCUCCCAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038092
          miRBase ID: ami-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-33-1
          Stem-loop Accession # MI0018854
          Stem-loop Sequence
          UGCUGGGUGGCAGCUGUAGUGCAUUGUAGUUGCAUUGCAUGUUCUAGCAGUAUUUGUGCAAUGUUUCCGCAGUGCAAUACAGAGGCAUACCGUAUCUCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021934
          miRBase ID: aca-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Stem-loop ID aca-mir-33-2
          Stem-loop Accession # MI0018855
          Stem-loop Sequence
          AGCUCAAGUGGCAGUUUCGGGGGUGCAUUGUAGUUGCAUUGCAUGUGUGGCCAAGCUGUGCAAUGCCCCUCCAGUGCAGCCCUGAGGCCGUCCUCUACCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021934
          miRBase ID: aca-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Anopheles gambiae

          Stem-loop ID aga-mir-33
          Stem-loop Accession # MI0033352
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCACUUACCGUGUUGGCAACAGGUGCAGUACUUCUGCAAUGCAACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041523
          miRBase ID: aga-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Bombyx mori

          Stem-loop ID bmo-mir-33
          Stem-loop Accession # MI0012293
          Stem-loop Sequence
          UAAAGCGAGAGUGCAUUGUAGUUGCAUUGCACCGGUUCCAUUAACGUGCAAUAUGACUACAAGGCAAAUCCCGAAUUAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013599
          miRBase ID: bmo-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-33a
          Stem-loop Accession # MI0009807
          Stem-loop Sequence
          CUGCGGUGCAUUGUAGUUGCAUUGCAUGUUCUGGCGGUACCCGUGCAAUGUUUCCACAGUGCAUCACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009294
          miRBase ID: bta-miR-33a
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Branchiostoma belcheri

          Stem-loop ID bbe-mir-33-1
          Stem-loop Accession # MI0026234
          Stem-loop Sequence
          AGACCUGUUGCUGGGGUGCAUUGUAGUUGCAUUGCAUGUGUGUUCAUUGCUGCAAUGUAACUGCAGUGCAGCCCAGAGGCAGGAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031668
          miRBase ID: bbe-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Stem-loop ID bbe-mir-33-2
          Stem-loop Accession # MI0026235
          Stem-loop Sequence
          GCUGAGGUGCAUUGUAGUUGCAUUGCAUGUGCAAAAGUGGAAGUGCAAUGUAACAGCAGUGCAGCUCAGACACGGUUGCAAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031668
          miRBase ID: bbe-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Capitella teleta

          Stem-loop ID cte-mir-33
          Stem-loop Accession # MI0010096
          Stem-loop Sequence
          GGAAUACGCUUUGCACAUUGGUGCAUUGUAGUUGCAUUGCAUGGAAGUUCAACCGGUGCAGUGCUCCUGCAGUGCAAUCAAUGUGAGGCGUCCUACUACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009553
          miRBase ID: cte-miR-33
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-33-1
          Stem-loop Accession # MI0029432
          Stem-loop Sequence
          GGGGGGCAGCUGUAGUGCAUUGUAGUUGCAUUGCAUGUUCUGGCAGUACUUGUGCAAUGUUUCUGCAGUGCAGUACAGAGGCACUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037689
          miRBase ID: cpi-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Stem-loop ID cpi-mir-33-2
          Stem-loop Accession # MI0029433
          Stem-loop Sequence
          AGUGGCCGCAGCUGUGGUGCAUUGUAGUUGCAUUGCAUGUGUGAGAACAGGUGUGCAAUGCCCCUGCAGUGCAGCCCAGAUGGGGCUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037689
          miRBase ID: cpi-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-33-1
          Stem-loop Accession # MI0029937
          Stem-loop Sequence
          CCUGGGUGGCAGCUGUAGUGCAUUGUAGUUGCAUUGCAUGUUGUGGCAGUAACUGUGCAAUGUUCCUGCAGUGCAGUACAGAGGCACUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038464
          miRBase ID: cli-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Stem-loop ID cli-mir-33-2
          Stem-loop Accession # MI0029938
          Stem-loop Sequence
          CCCGGCCCUCGCGGGGGUGCAUUGUAGUUGCAUUGCAUGUGUCAGACUGGGAGUGCAAUGCCCCUGCCAUGCAGCCCGGCGGGGGCCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038464
          miRBase ID: cli-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Culex quinquefasciatus

          Stem-loop ID cqu-mir-33
          Stem-loop Accession # MI0013620
          Stem-loop Sequence
          GGGUGCAUUGUAGUUGCAUUGCACGUUUUGCGUUCGAAAAGUCCAGUGCAGUACUUCUGCAAUGCAAACCAGGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014431
          miRBase ID: cqu-miR-33
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-33a
          Stem-loop Accession # MI0038942
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGUUCUGGCAGUACCCGUGCAAUGUGUCCACAGUGCAUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047571
          miRBase ID: dno-miR-33a-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Dinoponera quadriceps

          Stem-loop ID dqu-mir-33
          Stem-loop Accession # MI0037482
          Stem-loop Sequence
          UAUCUGUUACAAGUGUGCAUUGUAGUUGCAUUGCAUGAAAUAUAACUAUGCAAUACUUCUACAGUGCAACUCCUGUGGCAGGUCGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045665
          miRBase ID: dqu-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-33b-1
          Stem-loop Accession # MI0036074
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGUGUUUCUGCCGGGUGCAAUGUGUCUGCAGCGCAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044147
          miRBase ID: gmo-miR-33b-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Stem-loop ID gmo-mir-33b-2
          Stem-loop Accession # MI0037991
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCACGUCUGCUUUCACCGUGCAGUGUUUCCGCUCUGCAACG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044147
          miRBase ID: gmo-miR-33b-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-33a
          Stem-loop Accession # MI0038598
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGUUCUGGCAGUACCGUGCAAUGUUUCCACAGUGCAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046918
          miRBase ID: cpo-miR-33a-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-33a
          Stem-loop Accession # MI0012935
          Stem-loop Sequence
          CUGUGGUGCAUUGUAGUUGCAUUGCAUGUUCUGGCGGUACCCAUGCAAUGUUUCCACAGUGCAUCACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013189
          miRBase ID: eca-miR-33a
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Manduca sexta

          Stem-loop ID mse-mir-33
          Stem-loop Accession # MI0020978
          Stem-loop Sequence
          GUUAUGACGGACUAUUGCGGGAGUGCAUUGUAGUUGCAUUGCACCUGUUCGUAAUAAACGUGCAAUAUUACUACAAGGCAAAUCUCGAACUAGUUGCAUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024429
          miRBase ID: mse-miR-33
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-33
          Stem-loop Accession # MI0012511
          Stem-loop Sequence
          GGUGCAUUGUAGUUGCAUUGCACGUCGUGUCCGGGCCGUGCAAUGUUUCCACAGUGCAUCCCAGAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012754
          miRBase ID: mdo-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-33a
          Stem-loop Accession # MI0034762
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGAUCCUGAAGCCGAGUGCAAUGUGUCUGCAGUGCAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042819
          miRBase ID: oni-miR-33a-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Stem-loop ID oni-mir-33b
          Stem-loop Accession # MI0034679
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGUGUGUCUCAGCUGAGUGCAAUGUACCUGCAGUGCAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042753
          miRBase ID: oni-miR-33b-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-33a
          Stem-loop Accession # MI0032484
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGUUCUGGUGGUACCCAUGCAGUGUUUCCACAGUGCAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040081
          miRBase ID: pal-miR-33a-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-33-1
          Stem-loop Accession # MI0030191
          Stem-loop Sequence
          GGGUGGCAACUGUAGUGCAUUGUAGUUGCAUUGCAUGUUCUGGCAAUACUCAUGCAAUGUUUCUGCAGUGCAGUUCAGAGGCAUGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038882
          miRBase ID: pbv-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Stem-loop ID pbv-mir-33-2
          Stem-loop Accession # MI0030192
          Stem-loop Sequence
          CUAUGGCUGUGUCGGUGGUGCAUUGUAGUUGCAUUGCACGUGCAGGUGAAGAUGUGCAAUGCCCCUCCAGUGCAGCCCUGGGGCAGCAUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038882
          miRBase ID: pbv-miR-33-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-33a
          Stem-loop Accession # MI0039255
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGUUCUGGAAGUACCGUGCAAUGUUUCCACAGUGCAUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048165
          miRBase ID: ocu-miR-33a-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-33a-1
          Stem-loop Accession # MI0026690
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCACUGGGCUGCAGGAUCAUGCAAUGUGUCUGCAGUGCAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032618
          miRBase ID: ssa-miR-33a-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Stem-loop ID ssa-mir-33a-2
          Stem-loop Accession # MI0026691
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGAUCCUGCAGCCCAUUGCAAUGUGUCUGCAGUGCAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032618
          miRBase ID: ssa-miR-33a-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Stem-loop ID ssa-mir-33b
          Stem-loop Accession # MI0026692
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGUGUCUCAGCUGGGUGCAAUGUUCCUGCAGUGCAAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032620
          miRBase ID: ssa-miR-33b-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Triops cancriformis

          Stem-loop ID tcf-mir-33
          Stem-loop Accession # MI0033200
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGGUCGGUUUAAGAUGUGCAGUGUUUCUGCAGUGCAAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041358
          miRBase ID: tcf-miR-33
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-33a
          Stem-loop Accession # MI0031127
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGUCCUGACAGUACCAUGCAAUGUUUCCACAGUGCAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036468
          miRBase ID: tch-miR-33a-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-33a
          Stem-loop Accession # MI0038323
          Stem-loop Sequence
          GUGCAUUGUAGUUGCAUUGCAUGUGAUAUCGGUGAUGUGCAAUGUGCCUGCAGUGCAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046580
          miRBase ID: xla-miR-33a-5p
          Mature miRNA Sequence: GUGCAUUGUAGUUGCAUUGCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465263_pri
          TaqMan™ Pri-miRNA Assay : Mm03307171_pri
          TaqMan™ Pri-miRNA Assay : Hs03293451_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12410
          TaqMan™ MicroRNA Assay : 465396_mat
          TaqMan™ MicroRNA Assay : 002135
          mirVana® miRNA inhibitor : MH12410
          mirVana® miRNA mimic : MC12410
          TaqMan™ Advanced miRNA Assay : rno481363_mir
          TaqMan™ Advanced miRNA Assay : 478347_mir
          TaqMan™ Advanced miRNA Assay : mmu482743_mir


          miRBase Alias:

          Human: hsa-miR-33a (v17)
          Mouse: mmu-miR-33 (v17)
          Rat: rno-miR-33 (v18)
          Aeromonas caviae Ae398: aca-miR-33 (v19)
          Bombyx mori: bmo-miR-33 (v18)
          Monodelphis domestica: mdo-miR-33 (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline