Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs03303345_pri
Assay Name:
hsa-mir-1-1
Stem-loop ID:
hsa-mir-1-1
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.20: 62554306 - 62554376 [+] on Build GRCh38
Stem-loop Location:
Chr.20: 62554306 - 62554376 [+] on Build GRCh38
Mature miRNA ID:
hsa-miR-1-5p
hsa-miR-1-3p
Species:
Human
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Hs03303345_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
54
Chromosome Location:
Chr. 20 - 62554507 - 62554507 [+] on 62554507 Build GRCh38
Species:
Human
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Human
hsa-miR-1-5p
MIMAT0031892
Human
hsa-miR-1-3p
MIMAT0000416
Stem-loop Details
Human
Stem-loop ID
hsa-mir-1-1
Stem-loop Accession #
MI0000651
Stem-loop Sequence
UGGGAAACAUACUUCUUUAUAUGCCCAUAUGGACCUGCUAAGCUAUGGAAUGUAAAGAAGUAUGUAUCUCA
Chromosome Location
Chr. 20 - 62554306 - 62554376 [+] on Build GRCh38
Mature MicroRNA
miRBase Accession #: MIMAT0031892
miRBase ID: hsa-miR-1-5p
Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAU
Chromosome Location: Chr. 20 - 62554306 - 62554376 [+] on Build GRCh38
miRBase Accession #: MIMAT0000416
miRBase ID: hsa-miR-1-3p
Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUAU
Chromosome Location: Chr. 20 - 62554306 - 62554376 [+] on Build GRCh38
Gene Family ID
MIPF0000038,
mir-1
Distance to Amplicon
109 bases
Back To Top
More Information
Related Products
TaqMan
™
MicroRNA Assay :
475545_mat
mir
Vana
®
miRNA inhibitor :
MH24049
mir
Vana
®
miRNA mimic :
MC24049
TaqMan
™
Advanced miRNA Assay :
478726_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM10617
Ambion
®
Pre-miR
™
miRNA Precursor :
PM10617
TaqMan
™
MicroRNA Assay :
002222
mir
Vana
®
miRNA inhibitor :
MH10617
mir
Vana
®
miRNA mimic :
MC10617
TaqMan
™
Advanced miRNA Assay :
477820_mir
TaqMan
™
Advanced miRNA Assay :
mmu482914_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline