Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs03305585_pri
Assay Name:
hsa-mir-548l
Stem-loop ID:
hsa-mir-548l
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.11: 94466495 - 94466580 [-] on Build GRCh38
Stem-loop Location:
Chr.11: 94466495 - 94466580 [-] on Build GRCh38
Mature miRNA ID:
hsa-miR-548l
Species:
Human
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Hs03305585_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
71
Chromosome Location:
Chr. 11 - 94466094 - 94466094 [+] on 94466094 Build GRCh38
Species:
Human
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Human
hsa-miR-548l
MIMAT0005889
Stem-loop Details
Human
Stem-loop ID
hsa-mir-548l
Stem-loop Accession #
MI0006361
Stem-loop Sequence
UAUUAGGUUGGUGCAAAAGUAUUUGCGGGUUUUGUCGUAGAAAGUAAUGGCAAAAACUGCAGUUACUUGUGCACCAACCAAAUGCU
Chromosome Location
Chr. 11 - 94466495 - 94466580 [-] on Build GRCh38
Mature MicroRNA
miRBase Accession #: MIMAT0005889
miRBase ID: hsa-miR-548l
Mature miRNA Sequence: AAAAGUAUUUGCGGGUUUUGUC
Chromosome Location: Chr. 11 - 94466495 - 94466580 [-] on Build GRCh38
Gene Family ID
MIPF0000317,
mir-548
Distance to Amplicon
366 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM13139
Ambion
®
Pre-miR
™
miRNA Precursor :
PM13139
TaqMan
™
MicroRNA Assay :
002904
mir
Vana
®
miRNA inhibitor :
MH13139
mir
Vana
®
miRNA mimic :
MC13139
TaqMan
™
Advanced miRNA Assay :
479391_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline