Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04349573_pri
          Assay Name:
          hsa-mir-6836
          Stem-loop ID:
          hsa-mir-6836
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.7: 2257515 - 2257577 [-] on Build GRCh38
          Stem-loop Location:
          Chr.7: 2257515 - 2257577 [-] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-6836-3p hsa-miR-6836-5p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs04349573_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          67

          Chromosome Location:

          Chr. 7 - 2257727 - 2257727 [+] on 2257727 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-6836-3p MIMAT0027575
          Human hsa-miR-6836-5p MIMAT0027574

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-6836
          Stem-loop Accession # MI0022682
          Stem-loop Sequence
          GGCUCCGCAGGGCCCUGGCGCAGGCAUCCAGACAGCGGGCGAAUGCCUCCCCCGGCCCCGCAG
          Chromosome Location Chr. 7 - 2257515 - 2257577 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0027575
          miRBase ID: hsa-miR-6836-3p
          Mature miRNA Sequence: AUGCCUCCCCCGGCCCCGCAG
          Chromosome Location: Chr. 7 - 2257515 - 2257577 [-] on Build GRCh38

          miRBase Accession #: MIMAT0027574
          miRBase ID: hsa-miR-6836-5p
          Mature miRNA Sequence: CGCAGGGCCCUGGCGCAGGCAU
          Chromosome Location: Chr. 7 - 2257515 - 2257577 [-] on Build GRCh38

          Gene Family ID
          Distance to Amplicon 129 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM30262
          Ambion® Pre-miR™ miRNA Precursor : PM30262
          mirVana® miRNA inhibitor : MH30262
          mirVana® miRNA mimic : MC30262
          TaqMan™ Advanced miRNA Assay : 480434_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM29836
          Ambion® Pre-miR™ miRNA Precursor : PM29836
          TaqMan™ MicroRNA Assay : 467207_mat
          mirVana® miRNA inhibitor : MH29836
          mirVana® miRNA mimic : MC29836
          TaqMan™ Advanced miRNA Assay : 483151_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-l8rg4:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0