Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm03307588_pri
Assay Name:
mmu-mir-468
Stem-loop ID:
mmu-mir-468
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.6: 81896599 - 81896676 [-] on Build GRCm38
Stem-loop Location:
Chr.6: 81896599 - 81896676 [-] on Build GRCm38
Mature miRNA ID:
mmu-miR-468-5p
mmu-miR-468-3p
Species:
Mouse
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Mm03307588_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
64
Chromosome Location:
Chr. 6 - 81896292 - 81896292 [+] on 81896292 Build GRCm38
Species:
Mouse
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Mouse
mmu-miR-468-5p
MIMAT0022699
Mouse
mmu-miR-468-3p
MIMAT0002109
Stem-loop Details
Mouse
Stem-loop ID
mmu-mir-468
Stem-loop Accession #
MI0002403
Stem-loop Sequence
AUAAGAAACUUGGCGUGUCGUGACUGAUGUACUGAUAAGAAACUCAGUGUGAUAUGACUGAUGUGCGUGUGUCUGUCU
Chromosome Location
Chr. 6 - 81896599 - 81896676 [-] on Build GRCm38
Mature MicroRNA
miRBase Accession #: MIMAT0022699
miRBase ID: mmu-miR-468-5p
Mature miRNA Sequence: GACUGAUGUACUGAUAAGAAACUCAGU
Chromosome Location: Chr. 6 - 81896599 - 81896676 [-] on Build GRCm38
miRBase Accession #: MIMAT0002109
miRBase ID: mmu-miR-468-3p
Mature miRNA Sequence: UAUGACUGAUGUGCGUGUGUCUG
Chromosome Location: Chr. 6 - 81896599 - 81896676 [-] on Build GRCm38
Gene Family ID
Distance to Amplicon
274 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM23450
Ambion
®
Pre-miR
™
miRNA Precursor :
PM23450
TaqMan
™
MicroRNA Assay :
474253_mat
mir
Vana
®
miRNA inhibitor :
MH23450
mir
Vana
®
miRNA mimic :
MC23450
TaqMan
™
Advanced miRNA Assay :
mmu481850_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM10226
Ambion
®
Pre-miR
™
miRNA Precursor :
PM10226
TaqMan
™
MicroRNA Assay :
001085
mir
Vana
®
miRNA inhibitor :
MH10226
mir
Vana
®
miRNA mimic :
MC10226
TaqMan
™
Advanced miRNA Assay :
mmu481849_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline