Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04272618_pri
          Assay Name:
          mmu-mir-466h
          Stem-loop ID:
          mmu-mir-466g mmu-mir-466h mmu-mir-669o
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.2: 10514595 - 10514674 [+] on Build GRCm38
          Stem-loop Location:
          Chr.2: 10514595 - 10514674 [+] on Build GRCm38 Chr.2: 10514891 - 10514971 [+] on Build GRCm38 Chr.2: 10514300 - 10514395 [+] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-466g mmu-miR-466h-3p mmu-miR-466h-5p mmu-miR-669o-3p mmu-miR-669o-5p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm04272618_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          99

          Chromosome Location:

          Chr. 2 - 10515103 - 10515103 [-] on 10515103 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-466g MIMAT0004883
          Mouse mmu-miR-466h-3p MIMAT0017274
          Mouse mmu-miR-466h-5p MIMAT0004884
          Mouse mmu-miR-669o-3p MIMAT0017347
          Mouse mmu-miR-669o-5p MIMAT0009421

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-466g
          Stem-loop Accession # MI0005510
          Stem-loop Sequence
          UGUGUUUUCGUGUGUGUGCAUGUGGAUGUAUGUAUAUGAUUUUGCAUAUACAGACACAUGCACACACAUGCGGCACACAC
          Chromosome Location Chr. 2 - 10514595 - 10514674 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004883
          miRBase ID: mmu-miR-466g
          Mature miRNA Sequence: AUACAGACACAUGCACACACA
          Chromosome Location: Chr. 2 - 10514595 - 10514674 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 401 bases
          Stem-loop ID mmu-mir-466h
          Stem-loop Accession # MI0005511
          Stem-loop Sequence
          UGUAUAUUUGUGUGUGCAUGUGCUUGUGUGUAUGUGAAUAUAUAUAUCAUACGCACGCACACACACACACAAAUGCAAGCA
          Chromosome Location Chr. 2 - 10514891 - 10514971 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017274
          miRBase ID: mmu-miR-466h-3p
          Mature miRNA Sequence: UACGCACGCACACACACAC
          Chromosome Location: Chr. 2 - 10514891 - 10514971 [+] on Build GRCm38

          miRBase Accession #: MIMAT0004884
          miRBase ID: mmu-miR-466h-5p
          Mature miRNA Sequence: UGUGUGCAUGUGCUUGUGUGUA
          Chromosome Location: Chr. 2 - 10514891 - 10514971 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 104 bases
          Stem-loop ID mmu-mir-669o
          Stem-loop Accession # MI0009945
          Stem-loop Sequence
          UGCAUGUGCAUGUGUGUAUAGUUGUGUGUGCAUGUUUAUGUGUGUAUUUGAAUAUACAUAACAUACACACACACGUAUAAACACAAGCACACACAC
          Chromosome Location Chr. 2 - 10514300 - 10514395 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017347
          miRBase ID: mmu-miR-669o-3p
          Mature miRNA Sequence: ACAUAACAUACACACACACGUAU
          Chromosome Location: Chr. 2 - 10514300 - 10514395 [+] on Build GRCm38

          miRBase Accession #: MIMAT0009421
          miRBase ID: mmu-miR-669o-5p
          Mature miRNA Sequence: UAGUUGUGUGUGCAUGUUUAUGU
          Chromosome Location: Chr. 2 - 10514300 - 10514395 [+] on Build GRCm38

          Gene Family ID MIPF0000316, mir-467
          Distance to Amplicon 680 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM19954
          Ambion® Pre-miR™ miRNA Precursor : PM19954
          TaqMan™ MicroRNA Assay : 463964_mat
          mirVana® miRNA inhibitor : MH19954
          mirVana® miRNA mimic : MC19954
          Ambion® Anti-miR™ miRNA Inhibitor : AM15060
          Ambion® Pre-miR™ miRNA Precursor : PM15060
          TaqMan™ MicroRNA Assay : 121176_mat
          mirVana® miRNA inhibitor : MH15060
          mirVana® miRNA mimic : MC15060
          TaqMan™ Advanced miRNA Assay : mmu482832_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19731
          Ambion® Pre-miR™ miRNA Precursor : PM19731
          mirVana® miRNA inhibitor : MH19731
          mirVana® miRNA mimic : MC19731
          Ambion® Anti-miR™ miRNA Inhibitor : AM12941
          Ambion® Pre-miR™ miRNA Precursor : PM12941
          TaqMan™ MicroRNA Assay : 002516
          mirVana® miRNA inhibitor : MH12941
          mirVana® miRNA mimic : MC12941
          Ambion® Anti-miR™ miRNA Inhibitor : AM12523
          Ambion® Pre-miR™ miRNA Precursor : PM12523
          TaqMan™ MicroRNA Assay : 241015_mat
          mirVana® miRNA inhibitor : MH12523
          mirVana® miRNA mimic : MC12523
          TaqMan™ Advanced miRNA Assay : mmu482638_mir


          miRBase Alias:

          Mouse: mmu-mir-466g (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline