Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM12733
          Assay Name: hsa-miR-345-5p
          miRBase Accession Number: MI0000825
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100307859 - 100307956 [+] on Build GRCh38
          Mature miRNA Sequence: GCUGACUCCUAGUCCAGGGCUC
          Species: Human, Nomascus leucogenys, Pan troglodytes, Pongo pygmaeus
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM12733
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000189, mir-345

          Mature miRNA Sequence:

          GCUGACUCCUAGUCCAGGGCUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-345-5p MIMAT0000772
          Nomascus leucogenys nle-miR-345 MIMAT0049378
          Pan troglodytes ptr-miR-345 MIMAT0008111
          Pongo pygmaeus ppy-miR-345 MIMAT0015846

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-345
          Stem-loop Accession # MI0000825
          Stem-loop Sequence
          ACCCAAACCCUAGGUCUGCUGACUCCUAGUCCAGGGCUCGUGAUGGCUGGUGGGCCCUGAACGAGGGGUCUGGAGGCCUGGGUUUGAAUAUCGACAGC
          Chromosome Location Chr. 14 - 100307859 - 100307956 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000772
          miRBase ID: hsa-miR-345-5p
          Mature miRNA Sequence: GCUGACUCCUAGUCCAGGGCUC
          Chromosome Location: Chr. 14 - 100307859 - 100307956 [+] on Build GRCh38

          Nomascus leucogenys

          Stem-loop ID nle-mir-345
          Stem-loop Accession # MI0040116
          Stem-loop Sequence
          GCUGACUCCUAGUCCAGGGCUCGUGAUGGCUGGUGGGCCCUGAACGAGGGGUCUGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049378
          miRBase ID: nle-miR-345
          Mature miRNA Sequence: GCUGACUCCUAGUCCAGGGCUC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-345
          Stem-loop Accession # MI0008630
          Stem-loop Sequence
          CCCAAACCCUAGGUCUGCUGACUCCUAGUCCAGGGCUCGUGAUGGCUGGUGGGCCCUGAACGAGGGGUCUGGAGGCCUGGGUUUGAAUAUCGACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008111
          miRBase ID: ptr-miR-345
          Mature miRNA Sequence: GCUGACUCCUAGUCCAGGGCUC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-345
          Stem-loop Accession # MI0014906
          Stem-loop Sequence
          ACCCAAACCCUAGGUCUGCUGACUCCUAGUCCAGGGCUCGUGAUGGCUGGUGGGCCCUGAACGAGGGGUCUGGAGGCCUGGGUUUGAAUAUCGCCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015846
          miRBase ID: ppy-miR-345
          Mature miRNA Sequence: GCUGACUCCUAGUCCAGGGCUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303659_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12733
          TaqMan™ MicroRNA Assay : 002186
          mirVana® miRNA inhibitor : MH12733
          mirVana® miRNA mimic : MC12733
          TaqMan™ Advanced miRNA Assay : 478366_mir


          miRBase Alias:

          Human: hsa-miR-345 (v17)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-xq5gc:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0