Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • On-demand videos
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12733
          Assay Name: hsa-miR-345-5p
          miRBase Accession Number: MI0000825
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100307859 - 100307956 [+] on Build GRCh38
          Mature miRNA Sequence: GCUGACUCCUAGUCCAGGGCUC
          Species: Human, Nomascus leucogenys, Pan troglodytes, Pongo pygmaeus
          Product Type: mirVana® miRNA mimic
          Assay ID MC12733

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000189, mir-345

          Mature miRNA Sequence:

          GCUGACUCCUAGUCCAGGGCUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-345-5p MIMAT0000772
          Nomascus leucogenys nle-miR-345 MIMAT0049378
          Pan troglodytes ptr-miR-345 MIMAT0008111
          Pongo pygmaeus ppy-miR-345 MIMAT0015846

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-345
          Stem-loop Accession # MI0000825
          Stem-loop Sequence
          ACCCAAACCCUAGGUCUGCUGACUCCUAGUCCAGGGCUCGUGAUGGCUGGUGGGCCCUGAACGAGGGGUCUGGAGGCCUGGGUUUGAAUAUCGACAGC
          Chromosome Location Chr. 14 - 100307859 - 100307956 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000772
          miRBase ID: hsa-miR-345-5p
          Mature miRNA Sequence: GCUGACUCCUAGUCCAGGGCUC
          Chromosome Location: Chr. 14 - 100307859 - 100307956 [+] on Build GRCh38

          Nomascus leucogenys

          Stem-loop ID nle-mir-345
          Stem-loop Accession # MI0040116
          Stem-loop Sequence
          GCUGACUCCUAGUCCAGGGCUCGUGAUGGCUGGUGGGCCCUGAACGAGGGGUCUGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049378
          miRBase ID: nle-miR-345
          Mature miRNA Sequence: GCUGACUCCUAGUCCAGGGCUC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-345
          Stem-loop Accession # MI0008630
          Stem-loop Sequence
          CCCAAACCCUAGGUCUGCUGACUCCUAGUCCAGGGCUCGUGAUGGCUGGUGGGCCCUGAACGAGGGGUCUGGAGGCCUGGGUUUGAAUAUCGACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008111
          miRBase ID: ptr-miR-345
          Mature miRNA Sequence: GCUGACUCCUAGUCCAGGGCUC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-345
          Stem-loop Accession # MI0014906
          Stem-loop Sequence
          ACCCAAACCCUAGGUCUGCUGACUCCUAGUCCAGGGCUCGUGAUGGCUGGUGGGCCCUGAACGAGGGGUCUGGAGGCCUGGGUUUGAAUAUCGCCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015846
          miRBase ID: ppy-miR-345
          Mature miRNA Sequence: GCUGACUCCUAGUCCAGGGCUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303659_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12733
          Ambion® Pre-miR™ miRNA Precursor : PM12733
          TaqMan™ MicroRNA Assay : 002186
          mirVana® miRNA inhibitor : MH12733
          TaqMan™ Advanced miRNA Assay : 478366_mir


          miRBase Alias:

          Human: hsa-miR-345 (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-xq5gc:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0