Search
Search

| Assay Name: | hsa-miR-1250-5p |
|---|---|
| miRBase Accession Number: | MI0006385 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACGGUGCUGGAUGUGGCCUUU |
| Species: | Human, Pan troglodytes, Pongo pygmaeus |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000696, mir-1250 |
Mature miRNA Sequence: | ACGGUGCUGGAUGUGGCCUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1250-5p | MIMAT0005902 |
| Pan troglodytes | ptr-miR-1250 | MIMAT0007976 |
| Pongo pygmaeus | ppy-miR-1250 | MIMAT0016175 |