Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu478583_mir

          Assay Name:
          Stem-loop Accession Number: MI0000270
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 198858873 - 198858982 [-] on Build GRCh38
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Species: Human, Rat, Astatotilapia burtoni, Goat, Horse, Monodelphis domestica, Ophiophagus hannah, Pan troglodytes, Pongo pygmaeus, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu478583_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000007, mir-181

          Mature miRNA Sequence:

          AACAUUCAUUGCUGUCGGUGGGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-181b-5p MIMAT0000257
          Rat rno-miR-181b-5p MIMAT0000859
          Astatotilapia burtoni abu-miR-181c MIMAT0042253
          Goat chi-miR-181b-5p MIMAT0036001
          Horse eca-miR-181b MIMAT0013179
          Monodelphis domestica mdo-miR-181b-5p MIMAT0004155
          View More Rows
          Ophiophagus hannah oha-miR-181b-5p MIMAT0036762
          Pan troglodytes ptr-miR-181b MIMAT0002625
          Pongo pygmaeus ppy-miR-181b MIMAT0002628
          Tupaia chinensis tch-miR-181b-5p MIMAT0036543

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-181b-1
          Stem-loop Accession # MI0000270
          Stem-loop Sequence
          CCUGUGCAGAGAUUAUUUUUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location Chr. 1 - 198858873 - 198858982 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000257
          miRBase ID: hsa-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: Chr. 1 - 198858873 - 198858982 [-] on Build GRCh38

          Stem-loop ID hsa-mir-181b-2
          Stem-loop Accession # MI0000683
          Stem-loop Sequence
          CUGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location Chr. 9 - 124693710 - 124693798 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000257
          miRBase ID: hsa-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: Chr. 1 - 198858873 - 198858982 [-] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-181b-1
          Stem-loop Accession # MI0000926
          Stem-loop Sequence
          CCUGUGCAGAGAUGAUGUUUACAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUAGAAAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location Chr. 13 - 54952903 - 54953012 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000859
          miRBase ID: rno-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: Chr. 13 - 54952903 - 54953012 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-181b-2
          Stem-loop Accession # MI0000927
          Stem-loop Sequence
          UGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAAUGUCAACCAACUCACUGGUCAAUGAAUGCAAACUGCGGGCCAAAAA
          Chromosome Location Chr. 3 - 23151455 - 23151542 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000859
          miRBase ID: rno-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: Chr. 13 - 54952903 - 54953012 [+] on Build Rnor_6.0

          Astatotilapia burtoni

          Stem-loop ID abu-mir-181c
          Stem-loop Accession # MI0033987
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGCAGCAGAAUAACUCACUGAUCAAUGAAUGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042253
          miRBase ID: abu-miR-181c
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-181b
          Stem-loop Accession # MI0030650
          Stem-loop Sequence
          CUGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGACUUUGAAUCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036001
          miRBase ID: chi-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-181b
          Stem-loop Accession # MI0012921
          Stem-loop Sequence
          CUGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013179
          miRBase ID: eca-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-181b-1
          Stem-loop Accession # MI0005344
          Stem-loop Sequence
          AAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUUAACUGUAUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCACUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004155
          miRBase ID: mdo-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Stem-loop ID mdo-mir-181b-2
          Stem-loop Accession # MI0023753
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUUUGAUUCAAUCAACUCACUGAUCAAUGAAUGCAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004155
          miRBase ID: mdo-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Stem-loop ID mdo-mir-181b-3
          Stem-loop Accession # MI0023796
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGUGAGAUCUGAGGAAAAACUCACCGAUGGAUGAAUGUCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004155
          miRBase ID: mdo-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-181b
          Stem-loop Accession # MI0031378
          Stem-loop Sequence
          AAUAAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUCGGGUUCGGUCAGCCCUCUGGCUAAUGAAUGCAAACUGCGGAUCAAACAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036762
          miRBase ID: oha-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-181b-1
          Stem-loop Accession # MI0002934
          Stem-loop Sequence
          CCUGUGCAGAGAUUAUUUUUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002625
          miRBase ID: ptr-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Stem-loop ID ptr-mir-181b-2
          Stem-loop Accession # MI0008556
          Stem-loop Sequence
          UGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCGAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002625
          miRBase ID: ptr-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-181b
          Stem-loop Accession # MI0002936
          Stem-loop Sequence
          CCUGUGCAGAGAUUAUUGUUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002628
          miRBase ID: ppy-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Stem-loop ID ppy-mir-181b-2
          Stem-loop Accession # MI0014845
          Stem-loop Sequence
          CUGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCGAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002628
          miRBase ID: ppy-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-181b
          Stem-loop Accession # MI0031203
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036543
          miRBase ID: tch-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465090_pri
          TaqMan™ Pri-miRNA Assay : Rn03464621_pri
          TaqMan™ Pri-miRNA Assay : Hs03302963_pri
          TaqMan™ Pri-miRNA Assay : Hs03303356_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12442
          Ambion® Pre-miR™ miRNA Precursor : PM12442
          mirVana® miRNA inhibitor : MH12442
          mirVana® miRNA mimic : MC12442
          TaqMan™ Advanced miRNA Assay : rno478583_mir
          TaqMan™ Advanced miRNA Assay : 478583_mir


          miRBase Alias:

          Mouse: mmu-miR-181b-5p (v21)

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline